Skip to Content
Merck
All Photos(1)

Key Documents

403857

Sigma-Aldrich

Boron tribromide solution

1.0 M in heptane

Synonym(s):

Tribromoboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BBr3
CAS Number:
Molecular Weight:
250.52
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in heptane

bp

~90 °C

density

0.869 g/mL at 25 °C

SMILES string

BrB(Br)Br

InChI

1S/BBr3/c2-1(3)4

InChI key

ILAHWRKJUDSMFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Drug intermediate 6-nitro-L-DOPA
  • Luminescent polystyrene derivatives with sterically protected carbazolylborane moieties
  • High-quality boron-doped graphene via Wurtz-type reductive coupling reaction
  • Mercapto-(+)-methamphetamine haptens for synthesis of (+)-methamphetamine conjugate vaccines with improved epitope densities
  • Micrometer-sized organic molecule-DNA hybrid structures
  • Borane complexes via electrophilic aromatic borylation reactions
  • A 5-HT2C receptor agonist
  • Biphenyl-derivatives possessing tertiary amino groups as ß-secretase(BACE1) inhibitors for the treatment of Alzheimer′s disease
  • A highly near-IR region fluorescent p-extended boron aza-dipyrromethene moiety unit
  • Tetrahydroisoquinoline derivatives via intramolecular cyclization of methoxy-substituted N-phenethylimides

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hui Wang et al.
European journal of medicinal chemistry, 88, 66-73 (2014-09-14)
The pharmacokinetics (PK) and pharmacodynamics (PD) of PT119, a potent Staphylococcus aureus enoyl-ACP reductase (saFabI) inhibitor with a Ki value of 0.01 nM and a residence time of 750 min on the enzyme target, has been evaluated in mice. PT119
Andrey S Mereshchenko et al.
Nature chemistry, 7(7), 562-568 (2015-06-24)
'Roaming' is a new and unusual class of reaction mechanism that has recently been discovered in unimolecular dissociation reactions of isolated molecules in the gas phase. It is characterized by frustrated bond cleavage, after which the two incipient fragments 'roam'
Doriane Lorendeau et al.
Biochemical pharmacology, 90(3), 235-245 (2014-05-31)
The multidrug resistance protein 1 (MRP1) is involved in multidrug resistance of cancer cells by mediating drug efflux out of cells, often in co-transport with glutathione (GSH). GSH efflux mediated by MRP1 can be stimulated by verapamil. In cells overexpressing
Albish K Paul et al.
Photochemistry and photobiology, 91(6), 1348-1355 (2015-10-24)
Synthesis, photophysical and metal ion recognition properties of a series of amino acid-linked free-base and Zn-porphyrin derivatives (5-9) are reported. These porphyrin derivatives showed favorable photophysical properties including high molar extinction coefficients (>1 × 10(5) m(-1) cm(-1) for the Soret
Ho-Joong Kim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(33), 11836-11842 (2015-07-07)
Threefold symmetric rigid-core molecules with an internally grafted poly(ethylene oxide) (PEO) chain were synthesized, and their self-assembled structures were characterized using differential scanning calorimetry, TEM, and 1D and 2D X-ray scatterings in the solid state. The tripod compounds based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service