Skip to Content
Merck
All Photos(1)

Key Documents

433012

Sigma-Aldrich

Poly(dimethylsiloxane), vinyl terminated

average Mw ~25,000, viscosity 850-1,150 cSt (25 °C)(lit.)

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

mol wt

average Mw ~25,000
average Mw ~25,000

refractive index

n20/D 1.4052

viscosity

850-1,150 cSt(25 °C)(lit.)

bp

>93 °C (lit.)

iodine value

2.2‑3.5

density

0.965 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)=O

InChI

1S/C8H18OSi2/c1-7-10(3,4)9-11(5,6)8-2/h7-8H,1-2H2,3-6H3

InChI key

BITPLIXHRASDQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mold-making elastomers and encapsulants and sealants for electronic components.

Features and Benefits

Polymerized product provides non-corrosive shield against moisture, dirt and contaminants for electrical components. Polymer may be heat or room temperature cured. Polymerization produces no volatile gases.

Physical form

Divinyl functional reactive silicone fluid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. What is the weight of the product 433012-500ML in grams in a 500ml bottle?

    1 answer
    1. The weight of the product 433012-500ML in a 500ml bottle can be estimated by multiplying the volume (500ml) by the density (0.965 g/mL), resulting in an approximate weight of 482.5g in the bottle.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    0 answers

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service