Skip to Content
Merck
All Photos(1)

Key Documents

381802

Sigma-Aldrich

3,3′-Dimethoxybenzil

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCOC6H4OCH3
CAS Number:
Molecular Weight:
270.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

82-84 °C (lit.)

functional group

ketone

SMILES string

COc1cccc(c1)C(=O)C(=O)c2cccc(OC)c2

InChI

1S/C16H14O4/c1-19-13-7-3-5-11(9-13)15(17)16(18)12-6-4-8-14(10-12)20-2/h3-10H,1-2H3

InChI key

PJGXOGKIVAJFTE-UHFFFAOYSA-N

Gene Information

General description

3,3′-Dimethoxybenzil is a benzyl derivative. Modelling of the structured thermal diffuse scattering from 3,3′-dimethoxybenzil has been reported.[1]

Application

3,3′-Dimethoxybenzil may be used in the preparation of benzil derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational Disorder Enhances Solubility and Photovoltaic Performance of a Thiophene-Quinoxaline Copolymer.
Wang E, et al.
Advanced Energy Materials, 3(6), 806-814 (2013)
Modelling dynamic disorder in 3, 3'-dimethoxybenzil, C16H14O4.
Goossens DJ, et al.
Proceedings of the 16th Australian Institute of Physics Congress (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service