Skip to Content
Merck
All Photos(3)

Key Documents

B34729

Sigma-Aldrich

Biphenyl-4-carboxylic acid

95%

Synonym(s):

4-Phenylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CO2H
CAS Number:
Molecular Weight:
198.22
Beilstein:
973519
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

crystals

mp

220-225 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C13H10O2/c14-13(15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H,(H,14,15)

InChI key

NNJMFJSKMRYHSR-UHFFFAOYSA-N

Gene Information

human ... ALB(213) , BAD(572)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniil Naberezhnyi et al.
Physical chemistry chemical physics : PCCP, 22(17), 9808-9814 (2020-04-28)
Ionic liquids and nanoscale membranes are both considered as promising functional components to design next-generation gas separation technologies. Herein, we combine free-standing carbon nanomembranes (CNMs) with [bmim][Tf2N] ionic liquid having affinity to carbon dioxide, and explore molecular permeation through such
T Nanbo
Toxicology and applied pharmacology, 114(2), 302-305 (1992-06-01)
The relationship between the developmental change of the disposition kinetic character for p-phenylbenzoic acid (PPBA) and compartmentalized fluid of the body was investigated in the rat fetus. [14C]Inulin injected into the umbilical vein attained the steady state distribution at 8
T Nanbo
Journal of pharmacobio-dynamics, 5(4), 222-228 (1982-04-01)
The change in the pharmacokinetic character of p-phenyl benzoic acid (PPBA) in developing fetus of rat was investigated by a constant infusion of 14C-PPBA. PPBA level in tissues increased with the day of gestation. The hepatic clearance for PPBA showed
Pharmacokinetics of p-phenylbenzoic acid in near-term fetus of the rat.
T Nanbo
Chemical & pharmaceutical bulletin, 31(10), 3748-3751 (1983-10-01)
Kathrin Isenbügel et al.
Macromolecular rapid communications, 33(1), 41-46 (2011-10-26)
The evaporation driven self-assembly of novel colloidal silica Janus particles was evaluated by scanning electron microscopy in comparison to unfunctionalized silica particles. The cyclodextrin- and azobenzene-modified compound was obtained utilizing Pickering emulsion approach, in which the particles were immobilized on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service