Skip to Content
Merck
All Photos(3)

Key Documents

307696

Sigma-Aldrich

Cobalt(II) phthalocyanine

β-form, Dye content 97 %

Synonym(s):

Cobaltous phthalocyaninate, PcCo, Phthalocyanine cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16CoN8
CAS Number:
Molecular Weight:
571.46
Beilstein:
4121847
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

composition

Dye content, 97%

SMILES string

[Co]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Co/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

MPMSMUBQXQALQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) phthalocyanine (CoPC )is a metallopthalocyanine.

Application

Self assembled molecules of CoPC were deposited on epitaxial graphene. CoPC may be used to develop carbon black based electrocatalysts. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalysed aerobic regenerations of aldehydes and ketones have been investigated. CoPC may also be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of cyanide photocatalytic degradation using sol?gel ZnO sensitized with cobalt phthalocyanine
Guzman RRS, et al.
Journal of Sol-Gel Science and Technology, 54(4), 1-7 (2010)
Preparation and characterization of a novel hyperbranched poly(aryl ether ketone) terminated with cobalt phthalocyanine to be used for oxidative decomposition of 2,4,6-trichlorophenol
Lei L, et al.
Macromolecular Research, 18(4), 331-335 (2010)
Lin Cui et al.
Analytica chimica acta, 712, 64-71 (2011-12-20)
An enzyme-free amperometric method was established for the electrochemical reduction tert-butyl hydroperoxide (TBHP) on the utilization of nano-cobalt phthalocyanine (CoPc) loaded functionalized graphene (FGR) and to create a highly responsive organic peroxide sensor. FGR was synthesized with a simple and
Chicharro M, et al.
Analytical and Bioanalytical Chemistry, 331-335, 277-283 (2002)
Tatsuhiko Honda et al.
Journal of the American Chemical Society, 134(9), 4196-4206 (2012-02-04)
Proton-coupled electron-transfer reduction of dioxygen (O(2)) to afford hydrogen peroxide (H(2)O(2)) was investigated by using ferrocene derivatives as reductants and saddle-distorted (α-octaphenylphthalocyaninato)cobalt(II) (Co(II)(Ph(8)Pc)) as a catalyst under acidic conditions. The selective two-electron reduction of O(2) by dimethylferrocene (Me(2)Fc) and decamethylferrocene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service