Skip to Content
Merck
All Photos(1)

Documents

127221

Sigma-Aldrich

Di(2-pyridyl) ketone

99%

Synonym(s):

2,2′-Carbonyldipyridine, 2,2′-Dipyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
Beilstein:
126598
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

52-55 °C (lit.)

SMILES string

O=C(c1ccccn1)c2ccccn2

InChI

1S/C11H8N2O/c14-11(9-5-1-3-7-12-9)10-6-2-4-8-13-10/h1-8H

InChI key

QPOWUYJWCJRLEE-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a "star" topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes.

Biochem/physiol Actions

The Schiff base ligands derived from di-2-pyridyl ketone exhibit antiproliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Masternak et al.
Materials (Basel, Switzerland), 13(16) (2020-08-14)
The oncological use of cisplatin is hindered by its severe side effects and a very important resistance problem. To overcome these problems, scientists have attempted to design new generation transition-metal anticancer complexes. In this study, we present new complexes, ruthenium(II)
Hana Jansová et al.
Current medicinal chemistry, 26(2), 288-301 (2017-09-22)
Heart is a particularly sensitive organ to iron overload and cardiomyopathy due to the excessive cardiac iron deposition causes most deaths in disorders such as beta-thalassemia major. Free or loosely bound iron ions readily cycle between ferrous and ferric states
Maram T Basha et al.
Dalton transactions (Cambridge, England : 2003), 41(21), 6536-6548 (2012-03-01)
The iron coordination and biological chemistry of a series of heterocyclic dithiocarbazate Schiff base ligands is reported with regard to their activity as Fe chelators for the treatment of Fe overload and also cancer. The ligands are analogous to tridentate
Gergely Szakacs et al.
FEBS letters, 594(23), 3965-3985 (2020-10-25)
ABC transporters fulfill diverse physiological functions in different cellular localizations ranging from the plasma membrane to intracellular membranous compartments. Several ABC transporters have been spotted in the endolysosomal system, which consists of endosomes, autophagosomes, lysosomes, and lysosome-related organelles. In this
Constantinos G Efthymiou et al.
Dalton transactions (Cambridge, England : 2003), 39(37), 8603-8605 (2010-08-04)
Using monoanionic forms of di-2-pyridyl ketone as the only primary ligand, triangular Ni(2)M (M = lanthanide, Y) complexes with interesting magnetic properties have been synthesized.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service