Skip to Content
Merck
All Photos(2)

Documents

378801

Sigma-Aldrich

Calcium fluoride

random crystals, optical grade, 99.99% trace metals basis

Synonym(s):

Fluorite, Fluorspar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaF2
CAS Number:
Molecular Weight:
78.07
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050110
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

Assay

99.99% trace metals basis

form

random crystals

impurities

≤150.0 ppm Trace Metal Analysis

bp

2500 °C (lit.)

density

3.18 g/mL at 25 °C (lit.)

SMILES string

F[Ca]F

InChI

1S/Ca.2FH/h;2*1H/q+2;;/p-2

InChI key

WUKWITHWXAAZEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M ten Cate
European journal of oral sciences, 105(5 Pt 2), 461-465 (1997-12-12)
Low concentrations of fluoride have a beneficial effect on enamel and dentin de- and remineralization. After fluoride treatments, such as topical applications, rinses or dentifrices, salivary fluoride concentrations decrease exponentially in a biphasic manner to very low concentrations within a
Guanying Chen et al.
ACS nano, 6(9), 8280-8287 (2012-08-30)
We describe the development of novel and biocompatible core/shell (α-NaYbF(4):Tm(3+))/CaF(2) nanoparticles that exhibit highly efficient NIR(in)-NIR(out) upconversion (UC) for high contrast and deep bioimaging. When excited at ~980 nm, these nanoparticles emit photoluminescence (PL) peaked at ~800 nm. The quantum
Y Kitasako et al.
Journal of dental research, 91(4), 370-375 (2012-02-18)
The aim of this study was to assess the effect of chewing gum containing phosphoryl oligosaccharides of calcium (POs-Ca) and a low concentration of fluoride (F) on the hardness of enamel subsurface lesions, utilizing a double-blind, randomized, and controlled in
Nil Ozbek et al.
Food chemistry, 138(1), 650-654 (2012-12-26)
The determination of fluorine in milk samples via the molecular absorption of calcium mono-fluoride (CaF) was performed using a HR-CS-ETAAS. For this purpose, calcium was pipetted to graphite furnace together with samples. The amount of Ca and the graphite furnace
Lei Cheng et al.
Dental materials : official publication of the Academy of Dental Materials, 28(5), 573-583 (2012-02-10)
Previous studies have developed calcium phosphate and fluoride releasing composites. Other studies have incorporated chlorhexidine (CHX) particles into dental composites. However, CHX has not been incorporated in calcium phosphate and fluoride composites. The objectives of this study were to develop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service