Skip to Content
Merck
All Photos(2)

Key Documents

481521

Sigma-Aldrich

Hexaamminecobalt(III) chloride

99%

Synonym(s):

Cobalt hexammine trichloride, Hexaamminecobalt trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Co(NH3)6]Cl3
CAS Number:
Molecular Weight:
267.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
H2O: slightly soluble(lit.)
ethanol: insoluble(lit.)

Quality Level

Assay

99%

form

powder

mp

217 °C (lit.)

solubility

H2O: slightly soluble(lit.)
ethanol: insoluble(lit.)

density

1.71 g/mL at 25 °C (lit.)

SMILES string

N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]

InChI

1S/3ClH.Co.6H3N/h3*1H;;6*1H3/q;;;+3;;;;;;/p-3

InChI key

JXBGZYGSWFSYFI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hexaamminecobalt(III) chloride can be used as a starting material to prepare various Co-based nanomaterials such as Co-Sb nanoalloys, Co nanocapsules, bimetallic nanoparticles, and Co-polymer nanocomposites. These materials are applied in the field of catalysis, dielectrics, and renewable energy conversion devices.

Application

  • Rhenium (VII) compounds as inorganic precursors for the synthesis of organic reaction catalysts: Investigates the potential of Hexaamminecobalt(III) chloride as a replacement for commercial products in catalyzing organic reactions, highlighting its role in synthetic chemistry (K Leszczyńska-Sejda et al., 2019).
  • In situ formation of a molecular cobalt (iii)/AgCl photocatalyst for visible-light water oxidation: Discusses the synthesis and application of Hexaamminecobalt(III) chloride in forming a novel photocatalyst for water oxidation, a critical process in solar energy utilization (H Mogi et al., 2021).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bimetallic Pd-M (M?=?Pt, Ni, Cu, Co) nanoparticles catalysts with strong electrostatic metal-support interaction for hydrogenation of toluene and benzene
Jingyi Yang, et al.
Molecular Catalysis, 492, 110992-110992 (2020)
Sb-modulated synthesis of novel CoSb alloy nanoparticles anchored on N-doped carbon as oxygen electrocatalysts
Tao Gong, et al.
Applied Surface Science, 562, 150112-150112 (2021)
Synthesis of Co nanocapsules coated with BN layers by annealing of KBH4 and [Co(NH3)6]Cl3
Ichihito Narita, et al.
Solid State Communications, 137, 44-48 (2006)
Fabrication of a novel PANI/[Co (NH3)4 (C3H4N2)2]Cl3 nanocomposite with enhanced dielectric constant and ac-conductivity
Waseem Naqash and Kowsar Majid
Journal of Materials Science: Materials in Electronics, 28, 14217-14225 (2017)
Tumpa Sarkar et al.
Nucleic acids research, 33(1), 143-151 (2005-01-11)
The condensation of nucleic acids into well-defined particles is an integral part of several approaches to artificial cellular delivery. Improvements in the efficiency of nucleic acid delivery in vivo are important for the development of DNA- and RNA-based therapeutics. Presently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service