Skip to Content
Merck
All Photos(2)

Key Documents

378011

Sigma-Aldrich

Basic Blue 3

Dye content 25 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26ClN3O
CAS Number:
Molecular Weight:
359.89
Colour Index Number:
51004
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder or crystals

composition

Dye content, 25%

mp

205 °C (dec.) (lit.)

λmax

654 nm

ε (extinction coefficient)

≥12000 at 257-263 nm in water at 0.01 g/L
≥4500 at 289-295 nm in water at 0.01 g/L
≥45000 at 651-657 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CCN(CC)c1ccc2nc3ccc(cc3[o+]c2c1)N(CC)CC

InChI

1S/C20H26N3O.ClH/c1-5-22(6-2)15-9-11-17-19(13-15)24-20-14-16(23(7-3)8-4)10-12-18(20)21-17;/h9-14H,5-8H2,1-4H3;1H/q+1;/p-1

InChI key

IURGIPVDZKDLIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Basic blue 3 (BB3) can be removed from aqueous solutions with the help of activated carbon prepared from rubber seed coat.
Basic blue 3 (C.I. 51104) is a cationic dye. It is generally positively charged in an aqueous solution.

Application

Basic blue 3 has a brilliant blue shade and thus has been used as a colorant in dyeing acrylic fabrics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Ghanadzadeh Gilani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(1), 148-155 (2011-03-29)
The absorption and fluorescence spectra of a laser dye, oxazine-1 (OX1), in liquid and liquid crystalline media were studied at room temperature. The solvatochromic method was used to determine the ground- and excited-state dipole moments by means of Lippert-Mataga, Bakshiev
Miklós Kubinyi et al.
Applied spectroscopy, 59(1), 134-139 (2005-02-22)
It has been established by combined absorption and fluorescence measurements that the cationic dye Oxazine 1 (OX) and the polyvalent anionic host calix[8]arenesulfonate (SCA8) form two complexes in simultaneous reactions: OX + SCA8 <--> OX.SCA8 (1), and OX.SCA8 + OX
Umberto Giovanella et al.
Physical chemistry chemical physics : PCCP, 14(39), 13646-13650 (2012-09-12)
In a smart solution-processable luminescent poly(norbornene)/oxazine-1 (Ox1) intercalated fluoromica nanohybrid, the supramolecular organization of the Ox1 dyes can be tuned at the nanoscale level and a deep red emission band switched on by inducing a phase segregation of aligned molecules
A Ghanadzadeh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 73(2), 324-329 (2009-03-28)
The spectroscopic features of a laser dye, oxazine 1 (OX1), were investigated in liquid and in liquid crystalline solutions as a function of the solvent polarity and polarizability. The nature of spectral shift was investigated by studying the absorption spectra
H M Shapiro
Cytometry, 2(3), 143-150 (1981-11-01)
The addition of RNA content estimation to flow cytometric measurement of DNA content provides valuable information concerning cells' transitions between quiescent and proliferative states. Equilibrium staining methods employing acridine orange have been used for DNA/RNA content measurement but are difficult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service