Skip to Content
Merck
All Photos(1)

Key Documents

376035

Sigma-Aldrich

(Ethylthio)trimethylsilane

technical grade, 90%

Synonym(s):

Ethyl trimethylsilyl sulfide, Trimethylsilyl ethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SSi(CH3)3
CAS Number:
Molecular Weight:
134.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
90%

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.45 (lit.)

bp

69-70 °C/100 mmHg (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CCS[Si](C)(C)C

InChI

1S/C5H14SSi/c1-5-6-7(2,3)4/h5H2,1-4H3

InChI key

HXAFQWICVBBXMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(Ethylthio)trimethylsilane is reported to participate in the synthesis of adenophostin A and analog. It reacts with primary and secondary amines to yield aminosilane and ethanethiol.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some reactions of silyl-and germyl-sulphur compound with the S< sup> II</sup>? N bond.
Armitage DA and Clark MJ.
Journal of Organometallic Chemistry, 24(3), 629-631 (1970)
An expeditious route to the synthesis of adenophostin A.
van Straten NCR, et al.
Tetrahedron, 53(18), 6509-6522 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service