Skip to Content
Merck
All Photos(1)

Documents

194409

Sigma-Aldrich

Bromotrimethylsilane

97%

Synonym(s):

TMBS, Trimethylbromosilane, Trimethylsilyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiBr
CAS Number:
Molecular Weight:
153.09
Beilstein:
1731135
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.424 (lit.)

bp

79 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Br

InChI

1S/C3H9BrSi/c1-5(2,3)4/h1-3H3

InChI key

IYYIVELXUANFED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used with InCl3 to catalyze the direct allylation of a variety of alcohols with allyltrimethylsilane.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F

Flash Point(C)

32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Saito et al.
The Journal of organic chemistry, 71(22), 8516-8522 (2006-10-27)
The combination of InCl3 and Me3SiBr provided an enhanced Lewis acid system that can be used to promote a wide range of direct coupling reactions between alcohols and silyl nucleophiles in non-halogenated solvents, such as hexane or MeCN. The enhanced
Tetrahedron Letters, 34, 7713-7713 (1993)
Aldrichimica Acta, 14, 31-31 (1981)
M Nomizu et al.
International journal of peptide and protein research, 37(2), 145-152 (1991-02-01)
A new two-step deprotection/cleavage procedure for t-butoxycarbonyl (Boc) based solid phase peptide synthesis is reported. First the protective groups are removed from 4-(oxymethyl)-phenylacetamidomethyl (PAM) resin attached peptide with the weak hard acid, trimethylsilyl bromide-thioanisole/trifluoroacetic acid (TFA). In the second step
J T Sparrow et al.
Peptide research, 9(5), 218-222 (1996-09-01)
The original trimethylsilyl bromide (TMSBr) method of peptide resin deprotection and cleavage has been modified for convenience and applicability to larger peptides. Equal amounts of a 66-residue test peptide resin were cleaved by the standard hydrogen fluoride (HF) procedure, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service