Skip to Content
Merck
All Photos(1)

Documents

374784

Sigma-Aldrich

4′-Hydroxy-4-biphenylcarbonitrile

97%

Synonym(s):

4-Hydroxy-4′-cyanobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4CN
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid crystal

mp

191-195 °C (lit.)

SMILES string

Oc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C13H9NO/c14-9-10-1-3-11(4-2-10)12-5-7-13(15)8-6-12/h1-8,15H

InChI key

ZRMIETZFPZGBEB-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashmi Prabhu et al.
The journal of physical chemistry. B, 119(35), 11935-11952 (2015-08-12)
In this contribution, we present the results of our extensive investigations on the synthesis and phase behavior of five series of dimer-like compounds formed by covalently linking a promesogenic, cyanobiphenyl core, with a nonmesogenic N-(n-alkyl)-salicylaldimine segment via an oxy(oligomethylene)oxy spacer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service