Skip to Content
Merck
All Photos(1)

Documents

366854

Sigma-Aldrich

1-(trans-4-Hexylcyclohexyl)-4-isothiocyanatobenzene

liquid crystal (nematic), 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H10C6H4NCS
CAS Number:
Molecular Weight:
301.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid crystal (nematic)

refractive index

n20/D 1.532 (lit.)

density

0.977 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[C@H]1CC[C@@H](CC1)c2ccc(cc2)N=C=S

InChI

1S/C19H27NS/c1-2-3-4-5-6-16-7-9-17(10-8-16)18-11-13-19(14-12-18)20-15-21/h11-14,16-17H,2-10H2,1H3/t16-,17-

InChI key

STLICVZWECVJDT-QAQDUYKDSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ameer R K Nassrah et al.
Polymers, 13(2) (2021-01-13)
We provide experimental evidence for the influence of the molecular structure of the nematic liquid crystal (NLC) on the photoalignment process in three dimensions at the interface with a polymer layer. In particular, the experimental findings are explained through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service