374121
(1S)-(+)-3-Bromocamphor-10-sulfonic acid hydrate
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H15BrO4S · xH2O
CAS Number:
Molecular Weight:
311.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
optical activity
[α]20/D +94°, c = 1 in H2O
mp
117-121 °C (lit.)
functional group
bromo
ketone
sulfonic acid
SMILES string
O.CC1(C)[C@@H]2CC[C@@]1(CS(O)(=O)=O)C(=O)[C@H]2Br
InChI
1S/C10H15BrO4S.H2O/c1-9(2)6-3-4-10(9,5-16(13,14)15)8(12)7(6)11;/h6-7H,3-5H2,1-2H3,(H,13,14,15);1H2/t6-,7+,10-;/m1./s1
InChI key
RQZLIPHNDCFLPR-OFVIFIGMSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service