248940
(S)-(+)-1,1′-Binaphthyl-2,2′-diyl hydrogenphosphate
97%
Synonym(s):
(S)-(+)-1,1′-Binaphthalene-2,2′-diyl hydrogen phosphate, (S)-4-Hydroxydinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-oxide
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
optical activity
[α]22/D +595°, c = 1.35 in methanol
functional group
phosphate
SMILES string
OP1(=O)Oc2ccc3ccccc3c2-c4c(O1)ccc5ccccc45
InChI
1S/C20H13O4P/c21-25(22)23-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)24-25/h1-12H,(H,21,22)
InChI key
JEHUZVBIUCAMRZ-UHFFFAOYSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service