Skip to Content
Merck
All Photos(1)

Key Documents

359343

Sigma-Aldrich

Hydroquinidine

95%

Synonym(s):

(9S)-10,11-Dihydro-6′-methoxycinchonan-9-ol, Dihydroquinidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O2
CAS Number:
Molecular Weight:
326.43
Beilstein:
91441
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]25/D +226°, c = 2 in ethanol

mp

169-170 °C (lit.)

functional group

hydroxyl

SMILES string

CC[C@H]1CN2CCC1CC2[C@@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H26N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h4-6,8,11,13-14,19-20,23H,3,7,9-10,12H2,1-2H3/t13-,14-,19+,20-/m0/s1

InChI key

LJOQGZACKSYWCH-LHHVKLHASA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

General description

Hydroquinidine, a cinchona alkaloid, is a structural analog of quinidine. It shows antiarrhythmic effect.

Application

Hydroquinidine can be used as a chiral catalyst to prepare:
  • Enantioselective axially chiral phosphamides by N-allylic alkylation reaction between Morita-Baylis-Hillamn (MBH) carbonate and phosphamides.
  • Enantioselective α-amino phosphonates by hydrophosphonylation of N-aryl ketimines.
  • Enantioselective α-hydrazino esters by α-hydrazination of 3-oxoindolines using azodicarboxylate via aza-Michael reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of serum hydroquinidine determination by fluorescence polarization immunoassay and liquid chromatography.
Tod M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 8(3), 279-282 (1990)
Asymmetric Synthesis of Axially Chiral Phosphamides via Atroposelective N-Allylic Alkylation
Yang GH et al.
ACS Catalysis, 10, 2324-2333 (2020)
Catalytic enantioselective hydrophosphonylation of ketimines using cinchona alkaloids.
Nakamura S, et al.
Journal of the American Chemical Society, 131(51), 18240-18241 (2009)
Catalytic enantioselective hydrophosphonylation of ketimines using cinchona alkaloids
Nakamura, S, et al.
Journal of the American Chemical Society, 131, 18240-18241 (2009)
Relationships between heart rate variability and antiarrhythmic effects of hydroquinidine.
Brembilla-Perrot B and Jacquemin L
Cardiovascular Drugs and Therapy, 11(3), 493-498 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service