348805
Titanium
foil, thickness 0.5 mm, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
99.99% trace metals basis
form
foil
autoignition temp.
860 °F
resistivity
42.0 μΩ-cm, 20°C
thickness
0.5 mm
bp
3287 °C (lit.)
mp
1660 °C (lit.)
density
4.5 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
[Ti]
InChI
1S/Ti
InChI key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
General description
Ti has low thermal and electrical conductivity. It is highly corrosion-resistant and has a high strength to weight ratio. A few angstroms thick layer of titania on the surface of Ti products makes its corrosion resistant.5 Ti foils could be employed as a substrate to grow arrays of hematite nanorods by hydrothermal method. A study reports pressure less sintering of SiC pieces and single crystals to Ti foils at 1500oC. Ti foil may be applied as an interlayer, diffusion bonded during the self joining of Si3N4. Si3N4/Ti-foil/Si3N4.
Quantity
25×25 mm (approximately 1.4 g)
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Joining of silicon nitride with a titanium foil interlayer
Materials Science & Engineering. A, Structural Materials : Properties, Microstructure and Processing, 352, 169-178 (2003)
Bonding mechanism between silicon carbide and thin foils of reactive metals
J. Mater. Sci., 20(11), 3976-3982 (1985)
Large-Scale Porous Hematite Nanorod Arrays: Direct Growth on Titanium Foil and Reversible Lithium Storage.
The Journal of Physical Chemistry C, 114(49), 21158-21164 (2010)
International journal of computerized dentistry, 16(2), 125-141 (2013-08-13)
This article presents two novel options for lithium-disilicate restorations supported by single-tooth implants. By using a Ti-Base connector, hybrid abutments and hybrid abutment crowns can be fabricated for different implant systems. The latter option in particular is an interesting new
Journal of nanoscience and nanotechnology, 13(8), 5807-5810 (2013-07-26)
In this study, hydroxyapatite (HA) was coated on anodized titanium (Ti) surfaces through radio frequency magnetron sputtering in order to improve biological response of the titanium surface. All the samples were blasted with resorbable blasting media (RBM). RBM-blasted Ti surface
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service