Skip to Content
Merck
All Photos(1)

Key Documents

348864

Sigma-Aldrich

Titanium

wire, diam. 0.5 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
EC Number:
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

wire

autoignition temp.

860 °F

resistivity

42.0 μΩ-cm, 20°C

diam.

0.5 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

Application

  • Application and features of titanium for the aerospace industry: This article discusses the use of titanium in aerospace applications, highlighting its high strength-to-weight ratio and corrosion resistance, which are also critical in biomedical applications (Inagaki et al., 2014).
  • Opportunities and issues in the application of titanium alloys for aerospace components: This paper explores the engineering applications of titanium alloys, focusing on their performance in high-stress environments, which can translate to material science innovations (Williams & Boyer, 2020).
  • Titanium and titanium alloy applications in medicine: Discusses the applications of titanium in the medical field, particularly in implants due to its biocompatibility and strength, relevant to drug discovery and medical device fabrication (Jackson et al., 2016).
  • Titanium in biomedical applications—properties and fabrication: This review covers the properties of titanium that make it suitable for biomedical applications, including its use in prosthetics and implants, which is crucial for both material science and medical research (Jackson et al., 2016).
  • Titanium: An overview of resources and production methods: Provides a comprehensive overview of titanium sourcing and production techniques, essential for understanding material properties and applications in various industries (El Khalloufi et al., 2021).

Quantity

300 cm (approximately 2.7 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kurbad et al.
International journal of computerized dentistry, 16(2), 125-141 (2013-08-13)
This article presents two novel options for lithium-disilicate restorations supported by single-tooth implants. By using a Ti-Base connector, hybrid abutments and hybrid abutment crowns can be fabricated for different implant systems. The latter option in particular is an interesting new
Jinho Shin et al.
Journal of nanoscience and nanotechnology, 13(8), 5807-5810 (2013-07-26)
In this study, hydroxyapatite (HA) was coated on anodized titanium (Ti) surfaces through radio frequency magnetron sputtering in order to improve biological response of the titanium surface. All the samples were blasted with resorbable blasting media (RBM). RBM-blasted Ti surface
Study of a TiO2 photocatalytic coating for use in plasma catalysis.
K Van Wesenbeeck et al.
Communications in agricultural and applied biological sciences, 78(1), 227-233 (2013-07-24)
D M Rivera-Chacon et al.
Journal of biomedical nanotechnology, 9(6), 1092-1097 (2013-07-19)
Improvements in osteoconduction of implant biomaterials require focusing on the bone-implant interface, which is a complex multifactorial system. Surface topography of implants plays a crucial role at this interface. Nanostructured surfaces have been shown to promote serum protein adsorption and
Kwi-Dug Yun et al.
Journal of nanoscience and nanotechnology, 13(6), 3864-3867 (2013-07-19)
This study examined the bone response to titanium dioxide nanotube modified implants. A total of 24 implants were placed in the femur of 4 beagles. Before placement, screw-shaped implants were classified into 3 groups; machined surface (group M), titanium dioxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service