Skip to Content
Merck
All Photos(3)

Key Documents

346764

Sigma-Aldrich

Poly(vinyl chloride)

average Mw ~233,000, average Mn ~99,000

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mn ~99,000
average Mw ~233,000

inherent viscosity

1.40(lit.)

density

1.4 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher B Stipe et al.
Applied spectroscopy, 66(11), 1286-1293 (2012-11-14)
Airborne silica dust (quartz) is common in coal mines and represents a respiratory hazard that can lead to silicosis, a potentially fatal lung disease. With an eye toward developing a portable monitoring device for rapid analysis of silica dust, laser-induced
T D Ambrisko et al.
British journal of anaesthesia, 110(2), 305-310 (2012-11-21)
In a previous study, the authors found a large bias (50%) for lithium (LiDCO) compared with thermodilution cardiac output measurement methods in ponies receiving i.v. infusions of xylazine, ketamine, and midazolam. This prompted the authors to examine the effect of
Hitomi Sano et al.
PloS one, 7(11), e50212-e50212 (2012-12-05)
Although oxygen is essential for the wound healing process, tissue hypoxia is known to stimulate angiogenesis. To explore these inconsistent findings, we estimated the influence of the oxygen environment on wound healing with our original model. Experiment 1 (Establishment of
N A Soldatenko et al.
Gigiena i sanitariia, (1)(1), 42-46 (2013-06-29)
In the article the issues of environmental, sanitary and hygienic safety of medical waste management are considered. Recently, for the treatment of certain types of medical waste thermal methods using small plants not equipped with a proper flue gas cleaning
Jeffrey S Souris et al.
Molecular imaging, 12(1), 28-38 (2013-01-26)
At present, there is considerable interest in the use of in vivo fluorescence and bioluminescence imaging to track the onset and progression of pathologic processes in preclinical models of human disease. Optical quantitation of such phenomena, however, is often problematic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service