Skip to Content
Merck
All Photos(3)

Key Documents

189588

Sigma-Aldrich

Poly(vinyl chloride)

average Mw ~62,000, average Mn ~35,000

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

Quality Level

mol wt

average Mn ~35,000
average Mw ~62,000

inherent viscosity

0.68 dL/g(lit.)

density

1.4 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Neurotransmitter studies reagent in ventral hippocampal pathology: Eassa and colleagues (2023) explored the reversal of behavioral and neurophysiological correlates of psychosis in rats with ventral hippocampal Alzheimer′s disease-like pathology, potentially implicating neurotransmitter modulation by dopamine hydrochloride (Eassa et al., 2023).

  • Catecholamine research chemical in dopamine system dysfunction: McCoy et al. (2023) investigated the effects of a positive allosteric modulator of α5GABA(A) receptors on dopamine system dysfunction in temporal lobe epilepsy, where dopamine hydrochloride may serve as a critical reference material (McCoy et al., 2023).

  • Dopamine receptor agonist lab supply in GABA(A) receptor studies: Perez et al. (2023) studied how hippocampal α5-GABA(A) receptors modulate dopamine neuron activity in the rat ventral tegmental area, potentially employing dopamine hydrochloride as a research reagent (Perez et al., 2023).



Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Wang et al.
Environmental science & technology, 46(21), 11566-11574 (2012-10-11)
Opportunistic pathogens represent a unique challenge because they establish and grow within drinking water systems, yet the factors stimulating their proliferation are largely unknown. The purpose of this study was to examine the influence of pipe materials, disinfectant type, and
T D Ambrisko et al.
British journal of anaesthesia, 110(2), 305-310 (2012-11-21)
In a previous study, the authors found a large bias (50%) for lithium (LiDCO) compared with thermodilution cardiac output measurement methods in ponies receiving i.v. infusions of xylazine, ketamine, and midazolam. This prompted the authors to examine the effect of
Christopher B Stipe et al.
Applied spectroscopy, 66(11), 1286-1293 (2012-11-14)
Airborne silica dust (quartz) is common in coal mines and represents a respiratory hazard that can lead to silicosis, a potentially fatal lung disease. With an eye toward developing a portable monitoring device for rapid analysis of silica dust, laser-induced
Tariq Hussain et al.
International journal of pharmaceutics, 441(1-2), 781-789 (2012-10-16)
Electrostatic charge is generated during powder handling due to particle-particle and particle-wall collisions, rubbing, sliding, and rolling. In case of bipolar charge generation, the electrostatic forces may significantly change the inner forces and increase powder adhesion and cause a serious
Jeffrey S Souris et al.
Molecular imaging, 12(1), 28-38 (2013-01-26)
At present, there is considerable interest in the use of in vivo fluorescence and bioluminescence imaging to track the onset and progression of pathologic processes in preclinical models of human disease. Optical quantitation of such phenomena, however, is often problematic

Questions

  1. what is the refractive index of product_189588?

    1 answer
    1. Unfortunately, the refractive index is not determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service