Skip to Content
Merck
All Photos(1)

Key Documents

343854

Sigma-Aldrich

Methanol-d3

99.8 atom % D

Synonym(s):

1,1,1-Trideuteromethanol, Methyl-d3 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OH
CAS Number:
Molecular Weight:
35.06
Beilstein:
1732010
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.8 atom % D

Assay

99% (CP)

form

liquid

expl. lim.

5.5-36.5 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.05% water
water

refractive index

n20/D 1.327 (lit.)

bp

65 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])O

InChI

1S/CH4O/c1-2/h2H,1H3/i1D3

InChI key

OKKJLVBELUTLKV-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol-d3 is a deuterated NMR solvent that is useful in NMR-based research and analyses.

Application

Methanol-d3 may be used in the preparation of [Al(CH3)2OCD3]3 by reacting with trimethylaluminum for use in spectroscopic analysis of dialkylaluminum alkoxides and the trialkylaluminum-ether complex.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Władysław Wrzeszcz et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 349-354 (2017-07-29)
Recently, we demonstrated the presence of microheterogeneity in binary mixtures of unlike alcohols. [RSC Adv. 2016, 6, 37195-37202] The aim of this work was examination if this phenomenon occurs also in the mixture of very similar alcohols like CH
Thierry Jauffrais et al.
Marine drugs, 10(6), 1360-1382 (2012-07-24)
Azaspiracid (AZA) poisoning has been reported following consumption of contaminated shellfish, and is of human health concern. Hence, it is important to have sustainable amounts of the causative toxins available for toxicological studies and for instrument calibration in monitoring programs
Spectroscopic Studies of Dialkylaluminum Alkoxides and the Trialkyl-aluminum-Ether Complex. II. Assignments of Observed Frequencies
Tarao, R
Bulletin of the Chemical Society of Japan, 39(10), 2126-2132 (1966)
Paweł Tomza et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 197, 88-94 (2018-02-06)
Here we report ATR-IR spectroscopic study on the separation at a molecular level (microheterogeneity) and the degree of deviation of H2O/CH3OH and H2O/CD3OH mixtures from the ideal mixture. Of particular interest is the effect of isotopic substitution in methyl group
Jason S Adams et al.
Science (New York, N.Y.), 371(6529), 626-632 (2021-02-06)
Solvent molecules influence the reactions of molecular hydrogen and oxygen on palladium nanoparticles. Organic solvents activate to form reactive surface intermediates that mediate oxygen reduction through pathways distinct from reactions in pure water. Kinetic measurements and ab initio quantum chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service