Skip to Content
Merck
All Photos(2)

Key Documents

336807

Sigma-Aldrich

Iridium(III) chloride

crystalline

Synonym(s):

Iridium trichloride, Iridium(3+) chloride, Trichloroiridium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IrCl3
CAS Number:
Molecular Weight:
298.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline
Pricing and availability is not currently available.

grade

for analytical purposes

form

crystalline

concentration

35-37% (after oxygen combustion, silver nitrate titration)

mp

763 °C (lit.)

SMILES string

Cl[Ir](Cl)Cl

InChI

1S/3ClH.Ir/h3*1H;/q;;;+3/p-3

InChI key

DANYXEHCMQHDNX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajendra Prasad Paitandi et al.
European journal of medicinal chemistry, 84, 17-29 (2014-07-12)
Efficacy of the ferrocene appended piano-stool dipyrrinato complexes [(η(6)-C6H6)RuCl(fcdpm)] (1), [(η(6)-C10H14)RuCl(fcdpm)] (2), [(η(6)-C12H18)RuCl(fcdpm)] (3) [(η(5)-C5Me5)RhCl(fcdpm)] (4) and [(η(5)-C5Me5)IrCl(fcdpm)] (5) [fcdpm = 5-ferrocenyldipyrromethene] toward anticancer activity have been described. Binding of the complexes with calf thymus DNA (CT-DNA) and BSA (bovine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service