Skip to Content
Merck
All Photos(2)

Key Documents

307866

Sigma-Aldrich

Rhodium(III) chloride

98%

Synonym(s):

Rhodium trichloride, Rhodium(3+) trichloride, Trichlororhodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RhCl3
CAS Number:
Molecular Weight:
209.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
crystalline

Quality Level

Assay

98%

form

crystalline

reaction suitability

reagent type: catalyst
core: rhodium

SMILES string

Cl[Rh](Cl)Cl

InChI

1S/3ClH.Rh/h3*1H;/q;;;+3/p-3

InChI key

SONJTKJMTWTJCT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Rhodium(III) chloride precursor may be used to dope graphene with Rh nanoparticles to prepare hybrids for electrocatalysis in energy, sensing and biomedical sectors.

Physical form

Hydrophobic form

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leah C Matsinha et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 1240-1248 (2014-11-22)
A series of water-soluble Rh(i) mononuclear complexes of general formula: [Rh(sulphsal-X-R)(COD)] [sulphsal = sulphonated salicylaldimine, COD = cyclooctadiene; where R = H, Cl, CH3 and X = H, (t)Bu] have been synthesized. All the compounds were characterised using various spectroscopic
Keito Sano et al.
ACS applied materials & interfaces, 12(40), 44743-44753 (2020-09-12)
The molecular catalyst sensitized system (MCSS), where an excited molecular catalyst adsorbed on a semiconductor such as TiO2 injects electrons to the conduction band of the semiconductor leading to hydrogen evolution/CO2 reduction coupled with an oxidation of water on the
Noble metal (Pd, Ru, Rh, Pt, Au, Ag) doped graphene hybrids for electrocatalysis.
Marcella G, et al.
Nanoscale, 4(16), 5002-5008 (2012)
Yi Shi et al.
Nature communications, 11(1), 4558-4558 (2020-09-13)
The growth of atomically dispersed metal catalysts (ADMCs) remains a great challenge owing to the thermodynamically driven atom aggregation. Here we report a surface-limited electrodeposition technique that uses site-specific substrates for the rapid and room-temperature synthesis of ADMCs. We obtained
Polina Ivanova et al.
Nanotechnology, 32(5), 055704-055704 (2020-10-16)
Reactive oxygen species (ROS) play an important role in various physiological processes of living organisms. However, their increased concentration is usually considered as a threat for our health. Plants, invertebrates, and vertebrates including humans have various enzymatic and non-enzymatic defence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service