Skip to Content
Merck
All Photos(2)

Key Documents

328960

Sigma-Aldrich

Rose Bengal lactone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H4Cl4I4O5
CAS Number:
Molecular Weight:
973.67
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

95%

form

powder

mp

280 °C (dec.) (lit.)

λmax

557 nm

ε (extinction coefficient)

≥10000 at 317-323 nm
≥30000 at 516-522 nm
≥95000 at 554-560 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(I)cc2c(Oc3c(I)c(O)c(I)cc3C24OC(=O)c5c(Cl)c(Cl)c(Cl)c(Cl)c45)c1I

InChI

1S/C20H4Cl4I4O5/c21-9-7-8(10(22)12(24)11(9)23)20(33-19(7)31)3-1-5(25)15(29)13(27)17(3)32-18-4(20)2-6(26)16(30)14(18)28/h1-2,29-30H

InChI key

IICCLYANAQEHCI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ito et al.
Journal of the National Cancer Institute, 77(1), 277-281 (1986-07-01)
The tumorigenicity of 9-3',4',5',6'-tetrachloro-o-carboxy phenyl-6-hydroxy-2,4,5,7-tetraiodo-3-isoxanthone sodium (CAS: 632-68-8) [also called Food Red 105 (FR 105) or Rose Bengal B], which is widely used for food or cosmetic coloring in Japan, was examined in (C57BL/6N X C3H/N)F1 mice. Animals were divided
Y Hiasa et al.
Japanese journal of cancer research : Gann, 79(3), 314-319 (1988-03-01)
The effects of erythrosine (Red 3), rose bengal B (Red 105) and thyroidectomy on the development of thyroid tumor were examined in male Wistar rats treated with N-bis(2-hydroxypropyl)nitrosamine (DHPN). Red 3 and Red 105 were used at 4% in the
Iu D Shenin et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 41(2), 16-21 (1996-02-01)
The study was aimed at the development of a laboratory procedure for isomerization of polyene macrolides in the presence of sensitizers. The reaction was performed with exposure of levorin solutions to the xenon lamp irradiation in the presence of rose
D W Marshall et al.
Appetite, 39(3), 193-206 (2002-12-24)
Most people get married or spend time living with a partner at some stage in their lives. But what effect does this change have, if any, on their eating habits? The transition from bachelor or spinster to young couple represents
J Kanno et al.
Toxicology, 99(1-2), 107-113 (1995-05-05)
The effect of 3,4,5,6-tetrachloro-2',4',5',7'-tetraiodo-fluorescein sodium salt (Rose Bengal B or FD&C Red No. 105, molecular weight 1017.6) on the thyroid of rats treated with N-bis(2-hydroxypropyl)-nitrosamine (DHPN) and iodine-deficient (I-def) diet is studied. Six-week-old male F344 rats were divided into seven

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service