Skip to Content
Merck
All Photos(4)

Key Documents

E6003

Sigma-Aldrich

Eosin Y disodium salt

Dye content ≥85%, Powder

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, D&C;Red No. 22, Eosin yellowish

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Colour Index Number:
45380
Beilstein:
3586809
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Eosin Y disodium salt, Dye content ≥85 %

form

powder

Quality Level

composition

Dye content, ≥85%

color

brown

pH

6.55 (29 °C, 84 °F)

mp

255 — 270 °C (491 - 518 °F at 977.9 hPa)

solubility

H2O: 1 mg/mL

density

1.018 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Eosin Y disodium salt has been used for determining sperm viability. The swollen and stained spermatozoa were considered dead. In corneal epithelium, it has been used in the procedure of immunohistological staining.

Biochem/physiol Actions

Eosin Y is a xanthene dye and is used for the differential staining of connective tissue and cytoplasm. In histopathology, it is applied as a counterstain after hematoxylin and before methylene blue. It is also used as a background stain, thereby giving contrast to the nuclear stains.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

376.9 °F - Pensky-Martens closed cup

Flash Point(C)

191.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparison of lyophilized amniotic membrane with cryopreserved amniotic membrane for the reconstruction of rabbit corneal epithelium.
Ahn J, et al.
Biotechnology and Bioprocess Engineering: BBE, 10, 262-269 (2005)
Quality of sperm obtained by penile vibratory stimulation and percutaneous vasal sperm aspiration in men with spinal cord injury.
Qiu Y, et al.
Journal of Andrology, 33, 1036-1036 (2012)
Bancroft JD
Theory and Practice of Histological Techniques (2008)
Gangqi Wang et al.
Cells, 9(4) (2020-04-03)
We recently reported that loss of hyaluronan (HA) from the endothelial glycocalyx leads to loss of vessel stability in specific microcirculatory vascular beds. Here we hypothesized that such derangements in the glycocalyx may also impair the adaptive response to vascular
Histophatologic Techniques null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service