Skip to Content
Merck
All Photos(1)

Key Documents

323322

Sigma-Aldrich

Trimethylphosphine

97%

Synonym(s):

PMe3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3P
CAS Number:
Molecular Weight:
76.08
Beilstein:
969138
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7.24 psi ( 20 °C)

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Mitsunobu Reaction

reagent type: ligand
reaction type: Wittig Reaction

refractive index

n20/D 1.428 (lit.)

bp

38-40 °C (lit.)

mp

−86 °C (lit.)

density

0.738 g/mL at 20 °C (lit.)

functional group

phosphine

storage temp.

2-8°C

SMILES string

CP(C)C

InChI

1S/C3H9P/c1-4(2)3/h1-3H3

InChI key

YWWDBCBWQNCYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trimethylphosphine (PMe3) is an electron-rich phosphine ligand used as a reagent in Mitsunobu reaction.
It can be used:
  • In transformation of azides into carbamates; aziridines to azidoalcohols; and ketoximes to ketones with 2,2′-dipyridyl diselenide.
  • In Aza-Wittig reaction;, C-H bond activation of imines; cross-coupling reactions.
  • In the preparation of (cyanomethylene) trimethylphosphorane (CMMP) which is used as a reagent in Mitsunobu type reaction.
  • As a reagent in the synthesis of ruthenium trimethylphosphine complexes, which in turn are used as catalysts for hydrogenation of CO2 to formic acid.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthetic-technical development of oseltamivir phosphate [TM="Tamiflu"]: A race against time
Abrecht S, et al.
Chimia, 61(3), 93-99 (2007)
C- H Activation of Imines by Trimethylphosphine-Supported Iron Complexes and Their Reactivities
Camadanli S, et al.
Organometallics, 28(7), 2300-2310 (2009)
Hydrogenation of carbon dioxide catalyzed by ruthenium trimethylphosphine complexes: the accelerating effect of certain alcohols and amines
Munshi P, et al.
Journal of the American Chemical Society, 124(27), 7963-7971 (2002)
Synthesis of trifluoromethyl ketones by palladium-catalyzed cross-coupling reaction of phenyl trifluoroacetate with organoboron compounds
Kakino R, et al.
Bulletin of the Chemical Society of Japan, 74(2), 371-376 (2001)
One-pot conversion of azides to Boc-protected amines with trimethylphosphine and Boc-ON
Ariza X, et al.
Tetrahedron Letters, 39(49), 9101-9102 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service