Skip to Content
Merck
All Photos(1)

Key Documents

30881

Sigma-Aldrich

1-Butyl-3-methylimidazolium methanesulfonate

≥95%

Synonym(s):

1-Butyl-3-methyl-1H-imidazolium methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O3S
CAS Number:
Molecular Weight:
234.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

impurities

≤0.5% water

mp

75-80 °C (lit.)

anion traces

chloride (Cl-): ≤2%

functional group

sulfonic acid

SMILES string

CS([O-])(=O)=O.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.CH4O3S/c1-3-4-5-10-7-6-9(2)8-10;1-5(2,3)4/h6-8H,3-5H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

PUHVBRXUKOGSBC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-3-methylimidazolium methanesulfonate can be used:
  • To prepare a deep eutectic solvent applicable in the ammonia sorption.
  • As a solvent in the high-pressure CO2 removal process.
  • To prepare ionic liquid/phosphomolybdate based hybrids as catalysts applicable in the glucose epimerization.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

246.2 °F

Flash Point(C)

119 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Epimerization of glucose over ionic liquid/phosphomolybdate hybrids: structure-activity relationship
Meg'ias-Sayago C, et al.
Green Chemistry, 20(5), 1042-1049 (2018)
Evaluation of methanesulfonate-based deep eutectic solvent for ammonia sorption
Akhmetshina AI, et al.
Journal of Chemical and Engineering Data, 63(6), 1896-1904 (2018)
Ionic Liquid (1-Butyl-3-Metylimidazolium Methane Sulphonate) Corrosion and Energy Analysis for High Pressure CO2 Absorption Process
Taimoor AA, et al.
Processes, 6(5), 45-45 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service