Skip to Content
Merck
All Photos(1)

Key Documents

199982

Sigma-Aldrich

Tris(triphenylphosphine)rhodium(I) chloride

Synonym(s):

NSC 124140, RhCl(PPh3)3, Rhodium(I) tris(triphenylphosphine) chloride, Wilkinson’s catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]3RhCl
CAS Number:
Molecular Weight:
925.22
Beilstein:
4581440
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

Cl[Rh].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.ClH.Rh/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;1H;/q;;;;+1/p-1

InChI key

IXAYKDDZKIZSPV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tris(triphenylphosphine)rhodium(I) chloride is used as a decarbonylation reagent and as a cocatalyst in Heck reaction.[1] [2]

Application

Hydrosilylation Catalysts

Catalyst used for many organic reactions including:
  • Chemoselective allylic alkylations
  • Stoichiometric activation of Si-H bonds and hydrosilylations
  • Inter- and intramolecular hydroacylation of alkenes along with a cocatalyst
  • Polymerization of diorganostannanes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium (II) acetate- Tris (triphenylphosphine) rhodium (I) chloride: a novel catalytic couple for the intramolecular heck reaction
Donald B
The Journal of Organic Chemistry, 64, 3461-3466 (1999)
Stereochemistry of tris (triphenylphosphine) rhodium chloride decarbonylation of aldehydes
HM Walborsky et al.
Journal of the American Chemical Society, 93, 5465-5468 (1971)
The Journal of Organic Chemistry, 57, 5075-5075 (1992)
Chemical Reviews, 91, 1179-1179 (1991)
Effect of catalytic hydrogenation of cellular lipid and fatty acid on the susceptibility of tumor cells to humoral immune killing.
S I Schlager
Methods in enzymology, 73(Pt B), 191-199 (1981-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service