Skip to Content
Merck
All Photos(1)

Key Documents

302473

Sigma-Aldrich

Chlorobis(cyclooctene)rhodium(I),dimer

98%

Synonym(s):

[Rh(coe)2Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H56Cl2Rh2
CAS Number:
Molecular Weight:
717.50
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

mp

190 °C (dec.) (lit.)

SMILES string

Cl[Rh].Cl[Rh].C1CCCC=CCC1.C2CCCC=CCC2.C3CCCC=CCC3.C4CCCC=CCC4

InChI

1S/4C8H14.2ClH.2Rh/c4*1-2-4-6-8-7-5-3-1;;;;/h4*1-2H,3-8H2;2*1H;;/q;;;;;;2*+1/p-2/b4*2-1-;;;;

InChI key

GQPAPAIPOLEZHT-XFCUKONHSA-L

Application

Chlorobis(cyclooctene)rhodium(I),dimer can be used as a catalyst:
  • To synthesize enantioenriched gem-diaryl sulfonates by asymmetric 1,4-addition of aryl boronic acids to α,β-unsaturated sulfonyl compounds.
  • In direct C-arylation of N-unprotected indoles and pyrroles.
  • In the carboxylation of arenes via C-H bond activation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral Phosphite-Olefin Ligands: Application in Rh-Catalyzed Asymmetric 1, 4-Addition of Arylboronic Acids to beta-Aryl-alpha, beta-unsaturated Sulfonates
Yue-Na Y and Ming-Hua X
Acta Chimica Sinica , 72(7), 815-819 (2014)
Direct C-arylation of free (NH)-indoles and pyrroles catalyzed by Ar- Rh (III) complexes assembled in situ
Wang X, et al.
Journal of the American Chemical Society, 127(14), 4996-4997 (2005)
Rhodium-catalyzed C-C bond cleavage reactions-an update
Korotvicka A, et al.
Current Organic Chemistry, 16(10), 1170-1214 (2012)
Substrate control of stereoselection in the rhodium (I) catalyzed intramolecular [4+ 2] cycloaddition reaction
O'Mahony DJR, et al.
Organic & Biomolecular Chemistry, 1(12), 2038-2040 (2003)
Fumitoshi Kakiuchi et al.
Accounts of chemical research, 35(10), 826-834 (2002-10-16)
The cleavage and addition of ortho C-H bonds in various aromatic compounds such as ketones, esters, imines, imidates, nitrile, and aldehydes to olefins and acetlylenes can be achieved catalytically with the aid of ruthenium catalysts. The reaction is generally highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service