Skip to Content
Merck
All Photos(4)

Documents

299685

Sigma-Aldrich

Palladium(II) trifluoroacetate

97%

Synonym(s):

Pd(TFA)2, Trifluoroacetic acid palladium(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3COO)2Pd
CAS Number:
Molecular Weight:
332.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: C-H Activation

mp

~220 °C

SMILES string

FC(F)(F)C(=O)O[Pd]OC(=O)C(F)(F)F

InChI

1S/2C2HF3O2.Pd/c2*3-2(4,5)1(6)7;/h2*(H,6,7);/q;;+2/p-2

InChI key

PBDBXAQKXCXZCJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Palladium(II) trifluoroacetate is a catalyst used in Suzuki-Miyaura, Heck, and Stille cross-coupling reactions.

Application

Catalyst used for the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes.
Catalyzes the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates, which can then be converted to keto alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John E Bercaw et al.
The Journal of organic chemistry, 73(21), 8654-8657 (2008-09-30)
Molecular oxygen can replace sacrificial olefins as the hydrogen acceptor in the palladium trifluoroacetate catalyzed dehydrogenation of cyclohexene and related cyclic olefins into aromatics. One of the major drawbacks of the homogeneous system is the tendency of the palladium trifluoroacetate
Tetrahedron Letters, 25, 4187-4187 (1984)
Joshua S Dickstein et al.
Organic letters, 9(13), 2441-2444 (2007-06-05)
A palladium-catalyzed aromatic decarboxylation reaction has been developed. With electron-rich aromatic acids, the reaction proceeds efficiently under fairly mild conditions in good yields. The method was useful with complex functionalized substrates containing hindered carboxylic acids.
Keisuke Kato et al.
Angewandte Chemie (International ed. in English), 48(18), 3326-3328 (2009-02-05)
Boxing clever: Direct conversion of a terminal alkyne group into a beta-methoxyacrylate is realized with the aid of the bis(oxazoline) ligand (box). Acetyl and ketal protecting groups, free hydroxy groups, and acid-sensitive glycosidic bonds are not affected under the reaction
Palladium(II)-Catalyzed Direct ortho-C-H Acylation of Anilides by Oxidative Cross-Coupling with Aldehydes using tert-Butyl Hydroperoxide as Oxidant
Chan C, et al.
Advanced Synthesis & Catalysis, 353, 2999-3006 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service