Skip to Content
Merck
All Photos(1)

Key Documents

293407

Sigma-Aldrich

2-Quinoxalinecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O2
CAS Number:
Molecular Weight:
174.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

208 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cnc2ccccc2n1

InChI

1S/C9H6N2O2/c12-9(13)8-5-10-6-3-1-2-4-7(6)11-8/h1-5H,(H,12,13)

InChI key

UPUZGXILYFKSGE-UHFFFAOYSA-N

General description

Linear and Freundlich adsorption isotherm coefficient of 2-quinoxalinecarboxylic acid has been evaluated.

Application

2-Quinoxalinecarboxylic acid has been used in the preparation of:
  • N-(2-quinoxaloyl)-α-amino acids
  • bisquinoxaloyl (bisquinoxalinecarbonyl) derivatives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Prabavathi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 92, 325-335 (2012-03-27)
The FTIR and FT-Raman spectra of 2-quinoxaline carboxylic acid (2-QCA) has been recorded in the region 4000-450 and 4000-100 cm(-1), respectively. The conformational analysis, optimized geometry, frequency and intensity of the vibrational bands of 2-QCA were obtained by the density
M D Rose et al.
Food additives and contaminants, 12(2), 177-183 (1995-03-01)
A method for the determination of residues of quinoxaline-2-carboxylic acid (QCA), the major metabolite of carbadox, in pig kidney has been developed. Tissue samples were subjected to alkaline hydrolysis, liquid-liquid extractions, ion-exchange chromatography and further extraction to concentrate the analyte
Alex P Praseuth et al.
Biotechnology progress, 24(1), 134-139 (2008-01-05)
Proficient production of the antitumor agent triostin A was developed using engineered Escherichia coli (E. coli). The bacterium played host to 15 genes that encode integral biosynthetic proteins which were identified and cloned from Streptomyces lasaliensis. In this study, triostin
Joshua A Hagen et al.
Sensors (Basel, Switzerland), 11(7), 6645-6655 (2011-12-14)
Zinc oxide field effect transistors (ZnO-FET), covalently functionalized with single stranded DNA aptamers, provide a highly selective platform for label-free small molecule sensing. The nanostructured surface morphology of ZnO provides high sensitivity and room temperature deposition allows for a wide
Kento Koketsu et al.
Organic letters, 8(21), 4719-4722 (2006-10-06)
[reaction: see text] Little is known about how quinoxaline-2-carboxylic acid (QC) is synthesized in nature. On the basis of analysis of echinomycin biosynthetic gene clusters as well as feeding experiments with labeled precursors, we have proposed a biosynthetic pathway to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service