Skip to Content
Merck
All Photos(3)

Key Documents

200913

Sigma-Aldrich

Potassium hexafluorophosphate

≥99%

Synonym(s):

Monopotassium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KPF6
CAS Number:
Molecular Weight:
184.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

≥99%

form

solid

technique(s)

ion chromatography: suitable

mp

575 °C (lit.)

density

2.75 g/mL at 25 °C (lit.)

SMILES string

[K+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.K/c1-7(2,3,4,5)6;/q-1;+1

InChI key

YZDGRYDIGCWVND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium hexafluorophosphate can be used as an electrolyte additive to improve the:
  • Electrochemical performance of an anode-free lithium metal battery.
  • Cycling stability and Li Coulombic efficiency of lithium-sulfur(Li–S) batteries.
It can also be used as interface modification material to passivate interface defects in perovskite solar cells.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingru Li et al.
ACS applied materials & interfaces, 12(50), 56017-56026 (2020-12-04)
Uncontrollable dendrite growth and low Coulombic efficiency are the two main obstacles that hinder the application of rechargeable Li metal batteries. Here, an optimized amount of potassium hexafluorophosphate (KPF6, 0.01 M) has been added into the 2 M LiTFSI/ether-based electrolyte
Ruhamah Yunis et al.
Physical chemistry chemical physics : PCCP, 21(23), 12288-12300 (2019-05-30)
The synthesis and characterisation of new solid-state electrolytes is a key step in advancing the development of safer and more reliable electrochemical energy storage technologies. Organic ionic plastic crystals (OIPCs) are an increasingly promising class of material for application in
Yuexi Yang et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-02)
The use of baker's yeast to reduce ethyl 2-oxo-4-phenylbutyrate (EOPB) in conventional biphasic systems is hindered by low productivities due to mass transfer resistance between the biocatalyst and the substrate partitioned into two different phases. To overcome the limitation, a
Cong-Xiao Cao et al.
Scientific reports, 7(1), 3101-3101 (2017-06-10)
The unusually broad physical and chemical property window of ionic liquids allows for a wide range of applications, which gives rise to the recent spring-up of ionic liquid-based functional materials. Via solvothermal copolymerization of a monomeric ionic liquid and divinylbenzene
Ludmila Šimková et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 323, 106895-106895 (2021-01-12)
The molecule of 2,2-dinitroethene-1,1-diamine (FOX-7) is one of the most interesting molecules with multiple redox centres stabilized by push-pull effect. To reveal the detailed mechanism of its electrochemical process radical intermediates formed in the course of its electrochemical reduction in

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service