Skip to Content
Merck
All Photos(2)

Documents

291501

Sigma-Aldrich

Dichloro(chloromethyl)methylsilane

98%

Synonym(s):

(Chloromethyl)dichloromethylsilane, (Chloromethyl)methyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2)Si(CH3)Cl2
CAS Number:
Molecular Weight:
163.51
Beilstein:
969205
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

98%

refractive index

n20/D 1.449 (lit.)

bp

121-122 °C (lit.)

density

1.284 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](Cl)(Cl)CCl

InChI

1S/C2H5Cl3Si/c1-6(4,5)2-3/h2H2,1H3

InChI key

JAYBZWYBCUJLNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugen Weisheim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(35), 12436-12448 (2015-07-28)
Directed tridentate Lewis acids based on the 1,3,5-trisilacyclohexane skeleton with three ethynyl groups [CH2Si(Me)(C2H)]3 were synthesised and functionalised by hydroboration with HB(C6F5)2, yielding the ethenylborane {CH2Si(Me)[C2H2B(C6F5)2]}3, and by metalation with gallium and indium organyls affording {CH2Si(Me)[C2M(R)2]}3 (M = Ga, In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service