Skip to Content
Merck
All Photos(1)

Key Documents

290629

Sigma-Aldrich

N,N-Dimethylcyclohexylamine

99%

Synonym(s):

Cyclohexyldimethylamine, Dimethylaminocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N(CH3)2
CAS Number:
Molecular Weight:
127.23
Beilstein:
1919922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

3.6 mmHg ( 20 °C)

Quality Level

Assay

99%

autoignition temp.

419 °F

refractive index

n20/D 1.454 (lit.)

bp

158-159 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)C1CCCCC1

InChI

1S/C8H17N/c1-9(2)8-6-4-3-5-7-8/h8H,3-7H2,1-2H3

InChI key

SVYKKECYCPFKGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sampling and subsequent analytical determination of airborne N,N-dimethylcyclohexylamine has been reported.

Application

N,N-Dimethylcyclohexylamine has been used:
  • as switchable hydrophilicity solvent (SHS) for the extraction of lipids from freeze-dried samples of Botryococcus braunii microalgae for biofuel production
  • as catalyst in three-component organocatalyzed Strecker reaction on water

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillermo Lasarte-Aragonés et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-29)
In the present work, the effectiveness of switchable hydrophobicity solvents (SHSs) as extraction solvent (N,N-Dimethylcyclohexylamine (DMCA), N,N-Diethylethanamine (TEA), and N,N-Benzyldimethylamine (DMBA)) for a variety of emerging pollutants was evaluated. Different pharmaceutical products (nonsteroidal anti-inflammatory drugs (NSAIDs), hormones, and triclosan) were
[Toxic properties of dimethylcyclohexylamine].
E S Smirnova et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 54-55 (1984-05-01)
Fabio Cruz-Acosta et al.
Chemical communications (Cambridge, England), (44)(44), 6839-6841 (2009-11-04)
The first three-component organocatalyzed Strecker reaction operating on water has been developed. The manifold utilizes ketones (aldehydes) as the starting carbonyl component, aniline as the primary amine, acetyl cyanide as the cyanide source and N,N-dimethylcyclohexylamine as the catalyst.
M R Fitzpatrick et al.
American Industrial Hygiene Association journal, 44(6), 425-427 (1983-06-01)
A method has been developed for the collection on a reactive surface and subsequent analytical determination of airborne N,N-dimethylcyclohexylamine (DMCA). DMCA from ambient or test atmosphere is collected on a Tenax sorbent sample tube treated with hydrochloric acid (HCl) using
Xin Di et al.
Analytical and bioanalytical chemistry, 411(4), 803-812 (2018-12-14)
A simple and efficient method combining solid-phase extraction (SPE) with homogeneous liquid-liquid microextraction (HLLME) has been developed for fast pretreatment of chloramphenicol (CAP) from water samples prior to determination by high-performance liquid chromatography-ultraviolet detection. Oasis HLB sorbent was chosen for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service