Skip to Content
Merck
All Photos(1)

Key Documents

290572

Sigma-Aldrich

Tris(pentafluorophenyl)phosphine

97%

Synonym(s):

Tris(perfluorophenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6F5)3P
CAS Number:
Molecular Weight:
532.14
Beilstein:
773243
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

108-110 °C (lit.)

functional group

phosphine

SMILES string

Fc1c(F)c(F)c(P(c2c(F)c(F)c(F)c(F)c2F)c3c(F)c(F)c(F)c(F)c3F)c(F)c1F

InChI

1S/C18F15P/c19-1-4(22)10(28)16(11(29)5(1)23)34(17-12(30)6(24)2(20)7(25)13(17)31)18-14(32)8(26)3(21)9(27)15(18)33

InChI key

FQLSDFNKTNBQLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Chiavarino et al.
European journal of mass spectrometry (Chichester, England), 16(3), 407-414 (2008-01-01)
Functional models of the Compound I intermediate of monooxygenase heme enzymes, namely [(TPFPP)(*+)Fe(IV)=O](+) and [(TPFPP)Mn(V)=O](+) (TPFPP = meso-tetrakis (pentafluorophenyl)porphyrinato dianion), are obtained as bare species by electrospray ionization from solutions of appropriate precursors and their reactivity is investigated in the
A Karipides et al.
Acta crystallographica. Section C, Crystal structure communications, 45 ( Pt 11), 1743-1745 (1989-11-15)
C18F15P, Mr = 532.1, monoclinic, P2(1)/c, a = 7.194 (2), b = 17.930 (4), c = 13.834 (2) A, beta = 94.29 (2) degrees, V = 1779 A3, Z = 4, Dm = 2.00, Dx = 1.99 g cm-3, lambda(Mo
Rui Zhang et al.
Journal of the American Chemical Society, 127(18), 6573-6582 (2005-05-05)
Porphyrin-manganese(V)-oxo and porphyrin-manganese(IV)-oxo species were produced in organic solvents by laser flash photolysis (LFP) of the corresponding porphyrin-manganese(III) perchlorate and chlorate complexes, respectively, permitting direct kinetic studies. The porphyrin systems studied were 5,10,15,20-tetraphenylporphyrin (TPP), 5,10,15,20-tetrakis(pentafluorophenyl)porphyrin (TPFPP), and 5,10,15,20-tetrakis(4-methylpyridinium)porphyrin (TMPyP). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service