Skip to Content
Merck
All Photos(1)

Key Documents

283282

Sigma-Aldrich

Di(propylene glycol) methyl ether, mixture of isomers

Di(propylene glycol) methyl ether, mixture of isomers
1 of 1 reviewers received a sample product or took part in a promotion

97%

Synonym(s):

Dipropylene glycol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC3H6OC3H6OH
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

0.4 mmHg ( 25 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.422 (lit.)

bp

190 °C (lit.)

density

0.938 g/mL at 25 °C

SMILES string

O(C(CO)C)CC(OC)C

InChI

1S/C7H16O3/c1-6(4-8)10-5-7(2)9-3/h6-8H,4-5H2,1-3H3

InChI key

CUDYYMUUJHLCGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Koontz et al.
International journal of toxicology, 25(2), 95-107 (2006-04-07)
Computer modeling of aggregate exposure provides the capability to estimate the range of doses that can occur from product use and to understand the relative importance of different routes of exposure. This paper presents an assessment of aggregate occupational exposure
Toshiaki Tsukatani et al.
Chemical senses, 28(1), 25-32 (2002-12-28)
Detection thresholds are typically obtained by presenting a subject with serial dilutions of an odorant. Many factors, including the solvent used to dilute the odorant, can influence the measurement of detection thresholds. Differences have been reported in detection thresholds for
Minor pain inducers modulate membrane current in cultured neuronal cells.
K Inoue et al.
Pharmacology & toxicology, 77(6), 417-420 (1995-12-01)
R R Miller et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(4), 721-726 (1985-08-01)
Male Fischer 344 rats were given a single oral dose of approximately 1289 mg/kg (8.7 mmol/kg) of [14C]DPGME. After dosing, expired air, excreta, and tissues were analyzed for 14C activity, and metabolites in urine were isolated and identified. Approximately 60%
S Fairhurst et al.
Toxicology, 57(2), 209-215 (1989-07-17)
The subacute percutaneous toxicity of dipropylene glycol monomethyl ether (DPM) in male rats dosed 5 days/week for 4 weeks under both occluded and unoccluded conditions has been assessed and compared to the percutaneous toxicity of ethylene glycol monomethyl ether (EGM).

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. New Jersey
    • Reviews 3
    • Votes 0
    4 out of 5 stars.

    Great turnaround time & as specified.

    Good product turnaround & according to prescribed specs.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service