Skip to Content
Merck
All Photos(2)

Documents

279919

Sigma-Aldrich

2,7-Dimethylnaphthalene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
1852737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

263 °C (lit.)

mp

94-97 °C (lit.)

SMILES string

Cc1ccc2ccc(C)cc2c1

InChI

1S/C12H12/c1-9-3-5-11-6-4-10(2)8-12(11)7-9/h3-8H,1-2H3

InChI key

LRQYSMQNJLZKPS-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

The atmospheric oxidation mechanism of 2,7-dimethylnaphthalene initiated by OH radical was investigated. Adsorption of 2,7-dimethylnaphthalene isomers dissolved in supercritical carbon dioxide on NaY-type zeolite was also studied.

Application

2,7-Dimethylnaphthalene was used in the preparation of 2,7-bisbromomethylnapthalene via bromination with N-bromosuccinimide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of supercritical carbon dioxide+ 2, 6-and 2, 7-dimethylnaphthalene isomers on NaY-type zeolite.
Iwai Y, et al.
Industrial & Engineering Chemistry Research, 42(21), 5261-5267 (2003)
Zhijie Zhang et al.
The journal of physical chemistry. A, 117(1), 160-168 (2012-12-12)
The atmospheric oxidation mechanism of 2,7-dimethylnaphthalene (27DMN) initiated by OH radical is investigated at levels of BB1K and G3MP2-RAD//BH&HLYP. The reaction is mainly initiated by OH addition to the C(1) position to form radical adduct R1. In the atmosphere, R1
243. Eight-and higher-membered ring compounds. Part V. Di-(naphthalene-2: 7-dimethylene) and its conversion into coronene.
Baker W, et al.
Journal of the Chemical Society, 1118-1121 (1951)
Xu-Hui Huang et al.
Food & function, 12(4), 1626-1638 (2021-01-22)
Clam is a kind of nutritious, delicious and economical aquatic food around the world and is famous for its unique aroma. Instrumental analysis, sensory analysis, and comprehensive statistical analysis were performed to explain the relationship between aroma and odorants in
Kunal Roy et al.
European journal of medicinal chemistry, 44(5), 1941-1951 (2008-12-27)
A series of naphthalene and non-naphthalene derivatives (n=42) having cytochrome P450 2A6 and 2A5 inhibitory activities, reported by Rahnasto et al., were subjected to QSAR and QAAR studies. The analyses were performed using electronic, spatial, shape and thermodynamic descriptors to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service