Skip to Content
Merck
All Photos(2)

Key Documents

212806

Sigma-Aldrich

Diethylaluminum chloride solution

1.0 M in hexanes

Synonym(s):

Chlorodiethylaluminum, Dichlorotetraethyldialuminium, Diethylaluminum monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCl
CAS Number:
Molecular Weight:
120.56
Beilstein:
4123259
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in hexanes

density

0.711 g/mL at 25 °C

SMILES string

CC[Al](Cl)CC

InChI

1S/2C2H5.Al.ClH/c2*1-2;;/h2*1H2,2H3;;1H/q;;+1;/p-1

InChI key

YNLAOSYQHBDIKW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Diethylaluminum chloride solution can be used:
  • As a Lewis acid for the synthesis of Lewis acid-base bifunctional asymmetric catalysts for cyanosilylation of aldehydes and in Strecker reaction.
  • For the synthesis of precursors for lanthanide/aluminum catalysts for diene polymerization.
  • As a co-initiator in the cationic polymerization of isobutylene.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Catalytic asymmetric Strecker-Type Reaction Promoted by Lewis Acid-Lewis Base Bifunctional Catalvst.
Takamura M, et al.
Chemical & Pharmaceutical Bulletin, 48(10), 1586-1592 (2000)
Lanthanide carboxylate precursors for diene polymerization catalysis: Syntheses, structures, and reactivity with Et2AlCl.
Evans WJ, et al.
Organometallics, 20(26), 5751-5758 (2001)
Highly enantioselective cyanosilylation of aldehydes catalyzed by a Lewis acid-Lewis base bifunctional catalyst.
Hamashima Y, et al
Tetrahedron, 57(5), 805-814 (2001)
Synthesis of new graft copolymers by combining the DPE technique and cationic polymerization.
Wieland PC, et al.
Macromolecular Rapid Communications, 23(14), 809-813 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service