Skip to Content
Merck
All Photos(1)

Key Documents

274631

Sigma-Aldrich

1,2,2,6,6-Pentamethylpiperidine

97%

Synonym(s):

PMP, Pempidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21N
CAS Number:
Molecular Weight:
155.28
Beilstein:
103806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.460 (lit.)

bp

187-188 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CN1C(C)(C)CCCC1(C)C

InChI

1S/C10H21N/c1-9(2)7-6-8-10(3,4)11(9)5/h6-8H2,1-5H3

InChI key

XULIXFLCVXWHRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The focal 1,8-naphthalimide fluorophores in the antennae were modified with 1,2,2,6,6-pentamethylpiperidine to improve their photostability.

Application

1,2,2,6,6-Pentamethylpiperidine (PMP) was used as an organic structure directing agent (OSDA) in the synthesis of the RTH-type zeolites.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Maderdrut et al.
Brain research, 444(1), 189-194 (1988-03-15)
Both presynaptic and postsynaptic blockade of ganglionic transmission during the period of naturally occurring ganglion cell death reduced the number of surviving neurons in the sympathetic ganglia (SG) and ciliary ganglion (CG). The CG was chosen for analysis because there
C Maccarrone et al.
Life sciences, 42(25), 2633-2638 (1988-01-01)
Decentralization of the superior cervical ganglion (S.C.G.) of the rat elevated the neuropeptide-Y immunoreactivity (NPY-ir) content of the ganglion on day 1 (+43%) but not on day 3 post-surgery. The content of NPY-ir in the iris was increased by decentralization
[Tuberculous osteoarthritis revealed by ankle injury].
O Louletzian et al.
Presse medicale (Paris, France : 1983), 28(10), 531-532 (1999-04-21)
Nikolai I Georgiev et al.
Journal of fluorescence, 21(1), 51-63 (2010-07-20)
This paper reports on the divergent synthesis, photophysical properties and photodegradation of novel PAMAM dendrons, core and peripherally functionalized with 1,8-naphthalimide fluorophores. The novel compounds were configured as light-harvesting antennae where the system surface is labeled with blue emitting 4-allyloxy-1,8-naphthalimide
D Blozovski
Developmental psychobiology, 18(4), 355-366 (1985-07-01)
Young rats, 11, 16, and 20 days of age, received bilateral injections of three antinicotinic agents into the posteroventral hippocampo-subiculo-entorhinal area, and were trained to learn a cool-draft-stimulus, passive-avoidance task shortly after (17 min). Gallamine triethiodide had no action at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service