Skip to Content
Merck
All Photos(2)

Key Documents

270032

Sigma-Aldrich

N,N′-Dimethylethylenediamine

85%

Synonym(s):

1,2-Bis(methylamino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2CH2NHCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
878142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

85%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

119 °C (lit.)

density

0.819 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CNCCNC

InChI

1S/C4H12N2/c1-5-3-4-6-2/h5-6H,3-4H2,1-2H3

InChI key

KVKFRMCSXWQSNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N,N′-Dimethylethylenediamine was used to enhance CO2 adsorption. It was incorporated into H3[(Cu4Cl)3(BTTri)8 (CuBTTri; H3BTTri = 1,3,5-tri(1H-1,2,3-triazol-4-yl)benzene), a water-stable, triazolate-bridged framework, to form a metal–organic framework for CO2 separation.

Other Notes

remainder N-Methylethylenediamine

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced carbon dioxide capture upon incorporation of N, N?-dimethylethylenediamine in the metal?organic framework CuBTTri.
McDonald TM, et al.
Chemical Science, 2(10), 2022-2028 (2011)
Chandi C Malakar et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(29), 8882-8885 (2012-06-26)
Water makes it possible: The Cu(2)O-catalyzed reaction between easily available o-bromobenzylbromides and benzamidines by using Cs(2)CO(3) as the base and N,N'-dimethylethylenediamine (DMEDA) as the additive in water as the solvent gives access to substituted quinazolines in a single step with
Xiaohu Deng et al.
The Journal of organic chemistry, 74(15), 5742-5745 (2009-06-17)
CuI/L5 (N,N'-dimethylethylenediamine) proves to be an efficient catalyst system for the amination of arylhalides with guanidines. The same catalyst system is then successfully applied to the one-step synthesis of 1-H-2-amino-benzimidazoles through tandem aminations of 1,2-dihaloarenes in modest yields. This methodology
Mohamed R Shehata et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 383-388 (2012-03-06)
Pd(dmen)Cl(2) complex was synthesized and characterized, where dmen=N,N-dimethylethylenediamine. Stoichiometry and stability constants of the complexes formed between [Pd(dmen)(H(2)O)(2)](2+) and various biologically relevant ligands such as amino acids, peptides and dicarboxylic acids are investigated at 25 °C and at constant 0.1M
A Szent-Györgyi
Physiological chemistry and physics, 12(2), 99-110 (1980-01-01)
Most inanimate systems are build of closed-shell molecules in which electrons lack excitability and mobility. These electrons can be rendered reactive and mobile by taking out some of them, desaturating the system electronically. Single electrons can be taken out of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service