Skip to Content
Merck
All Photos(3)

Documents

268674

Sigma-Aldrich

2-Aminoethylphosphonic acid

99%

Synonym(s):

2-AEP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
125.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

296 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless

SMILES string

NCCP(O)(O)=O

InChI

1S/C2H8NO3P/c3-1-2-7(4,5)6/h1-3H2,(H2,4,5,6)

InChI key

QQVDJLLNRSOCEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The surface of gold nanoparticles (Au NPs) were functionalized with 2-aminoethylphosphonic acid that exhibited calcium affinity which enabled targeted delivery of Au NPs to calcified tissue.

Application

2-Aminoethylphosphonic acid was used as a growth medium for the marine bacterium Roseovarius nubinhibens ISM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhiqiang Xu et al.
Chemical communications (Cambridge, England), (7)(7), 878-879 (2003-05-13)
Carbon-supported catalysts were phosphonated using 2-aminoethylphosphonic acid, and the resulting catalysts with largely enhanced proton conductivity performed substantially better than the untreated counterparts in proton-exchange membrane fuel cells.
Celia C H Chen et al.
Biochemistry, 41(44), 13162-13169 (2002-10-31)
Phosphonates allow certain organisms to thrive in otherwise hostile environments, and 2-aminoethylphosphonate (AEP) is a precursor of many cellular phosphonates. AEP transaminase (AEPT) is an enzyme essential to phosphonate synthesis and degradation pathways. The crystal structure of AEP transaminase was
Magdalena Klimek-Ochab et al.
Biodegradation, 18(2), 223-231 (2006-06-08)
Air-born mixed fungal and bacterial culture capable of complete degradation of ciliatine was isolated. The utilization of the natural organophosphonate proceeded in the phosphate independent manner. Enzymatic activity involved in ciliatine degradation studied in the fungal cell-free extract proved to
Natalia V Zhukova
Lipids, 42(12), 1169-1175 (2007-10-26)
Two nudibranch mollusks, Chromodoris sp. and Phyllidia coelestis, collected from tropical waters of the Northwestern Pacific, were analyzed for lipids. The aim of this study was to fill the gap in knowledge of lipid biochemistry of mollusks. Phospholipids (PL) were
Mitali Sarkar et al.
The Journal of biological chemistry, 278(25), 22703-22708 (2003-04-04)
Phosphoenolpyruvate (PEP) mutase catalyzes the conversion of phosphoenolpyruvate to phosphonopyruvate, the initial step in the formation of many naturally occurring phosphonate compounds. The phosphonate compound 2-aminoethylphosphonate is present as a component of complex carbohydrates on the surface membrane of many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service