Skip to Content
Merck
All Photos(1)

Key Documents

268615

Sigma-Aldrich

3-Aminopropylphosphonic acid

98%

Synonym(s):

β-Aminopropylphosphonic acid, 3-Aminopropane-1-phosphonic acid, 3-Aminopropanephosphonic acid, 3-Azaniumylpropyl(hydroxy)phosphinate, P-(3-Aminopropyl)phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3P(O)(OH)2
CAS Number:
Molecular Weight:
139.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

294 °C (dec.) (lit.)

functional group

amine

SMILES string

NCCCP(O)(O)=O

InChI

1S/C3H10NO3P/c4-2-1-3-8(5,6)7/h1-4H2,(H2,5,6,7)

InChI key

GSZQTIFGANBTNF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The relative nonspecific binding of the DNA-conjugated gold nanoparticles (AuNPs) to the indium-tin oxide electrodes modified with 3-aminopropylphosphonic acid was examined by measuring the electrocatalytic anodic current of hydrazine caused by the nonspecifically absorbed AuNPs.

Application

3-Aminopropylphosphonic acid was used as a selective antagonist of GABA(C) receptor.

Biochem/physiol Actions

GABAB receptor agonist; antagonist of basal prolactin secretion.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Gillies et al.
The American journal of physiology, 267(1 Pt 1), C195-C203 (1994-07-01)
The extracellular pH (pHex) of tumors is generally acidic. However, it is only recently that noninvasive magnetic resonance spectroscopic (MRS) measurements have determined that the intracellular pH (pHin) of tumor cells in situ is neutral or slightly alkaline compared with
N V Kapousta-Bruneau
Vision research, 40(13), 1653-1665 (2000-05-18)
The largest component in the fully dark-adapted ERG is a corneal-positive response, known as the b-wave, and believed to originate from depolarizing (ON-type) bipolar cells. The two types of GABA receptors, GABA(A) and GABA(C) have been reported to exist on
New magnetic resonance imaging contrast agents for infarct imaging based on phosphonate derivatives of Gd-DTPA.
I K Adzamli et al.
Investigative radiology, 26 Suppl 1, S242-S244 (1991-11-01)
D I Kerr et al.
Brain research, 480(1-2), 312-316 (1989-02-20)
Baclofen, 3-amino-propylphosphonic acid (3-APPA), and beta-phenyl-GABA (BPG), each reduced the frequency of spontaneous paroxysmal discharges in rat neocortical slices maintained in Mg2+-free medium, reversibly antagonised by phaclofen and 4-amino-butylphosphonic acid (4-ABPA). At lower concentrations, not influencing the discharges, both 3-APPA
Robert J Gillies et al.
Annual review of biomedical engineering, 7, 287-326 (2005-07-12)
Magnetic resonance spectroscopy (MRS) has been used for more than two decades to interrogate metabolite distributions in living cells and tissues. Techniques have been developed that allow multiple spectra to be obtained simultaneously with individual volume elements as small as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service