Skip to Content
Merck
All Photos(1)

Key Documents

267724

Sigma-Aldrich

Zirconium

rod, diam. 6.35 mm, ≥99% trace metals basis

Synonym(s):

Zirconium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
EC Number:
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

rod

resistivity

40 μΩ-cm, 20°C

diam.

6.35 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

100 g = 50 cm; 20 g = 10 cm

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

O Tymofiyeva et al.
Dento maxillo facial radiology, 42(6), 20120271-20120271 (2013-04-24)
To investigate the potential influence of standard dental materials on dental MRI (dMRI) by estimating the magnetic susceptibility with the help of the MRI-based geometric distortion method and to classify the materials from the standpoint of dMRI. A series of
Surgical, biologic and implant-related factors affecting bone remodeling around implants.
Matteo Capelli
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry, 8(2), 279-313 (2013-05-29)
New CAD/CAM materials and blocks for chairside procedures.
M Zimmermann et al.
International journal of computerized dentistry, 16(2), 173-181 (2013-08-13)
[New bipolar electrosurgical tools based on zirconium dioxide].
S V Belov et al.
Meditsinskaia tekhnika, (2)(2), 20-24 (2013-06-26)
Gopalu Karunakaran et al.
Journal of nanoscience and nanotechnology, 13(1), 678-685 (2013-05-08)
Currently, nanometal oxides are used extensively in different industries such as medicine, cosmetics and food. The increased consumption of nanoparticles (NPs) leads the necessity to understand the fate of the nanoparticles in the environment. The present study focused on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service