Skip to Content
Merck
Zirconium foil, not light tested, 50x50mm, thickness 0.003mm, 99.2%

Zirconium

Zirconium foil, not light tested, 50x50mm, thickness 0.003mm, 99.2%
Synonym(s):
Zirconium
Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11