Skip to Content
Merck
All Photos(1)

Key Documents

264229

Sigma-Aldrich

N,N′-Bis(2,5-di-tert-butylphenyl)-3,4,9,10-perylenedicarboximide

Dye content 97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H50N2O4
CAS Number:
Molecular Weight:
766.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

composition

Dye content, 97%

mp

>300 °C (lit.)

λmax

528 nm

semiconductor properties

N-type (mobility=1.8x10−4 cm2/V·s)

SMILES string

CC(C)(C)c1ccc(c(c1)N2C(=O)c3ccc4c5ccc6C(=O)N(C(=O)c7ccc(c8ccc(C2=O)c3c48)c5c67)c9cc(ccc9C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C52H50N2O4/c1-49(2,3)27-13-23-37(51(7,8)9)39(25-27)53-45(55)33-19-15-29-31-17-21-35-44-36(22-18-32(42(31)44)30-16-20-34(46(53)56)43(33)41(29)30)48(58)54(47(35)57)40-26-28(50(4,5)6)14-24-38(40)52(10,11)12/h13-26H,1-12H3

InChI key

BIYPCKKQAHLMHG-UHFFFAOYSA-N

General description

N,N′-Bis(2,5-di-tert-butylphenyl)-3,4,9,10-perylenedicarboximide (BTBP) is a fluorescence dye that can be used as an electron acceptor. It has an extinction coefficient of 7.4 x 104 cm-1 mol-1 dm3.

Application

An n-channel organic semiconductor.
BTBP is a dye that can be used in the development of organic electronic based devices which include organic light emitting diodes(OLED), electroluminescent device and organic solar cells(OSCs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of doping dyes on the electroluminescent characteristics of multilayer organic light-emitting diodes.
Suzuki H and Hoshino S
Journal of Applied Physics, 79(11), 8816-8822 (1996)
The cybotactic region surrounding fluorescent probes dissolved in 1-butyl-3-methylimidazolium hexafluorophosphate: effects of temperature and added carbon dioxide.
Baker SN, et al.
The Journal of Physical Chemistry B, 105(39), 9663-9668 (2001)
Insulated molecular wire with highly conductive pi-conjugated polymer core.
Terao J, et al.
Journal of the American Chemical Society, 131(50), 18046-18047 (2009)
Self-assembled gelators for organic electronics.
Babu SS, et al.
Angewandte Chemie (International Edition in English), 51(8), 1766-1776 (2012)
Three-layered multicolor organic electroluminescent device.
Yoshida M, et al.
Applied Physics Letters, 69(6), 734-736 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service