Skip to Content
Merck
All Photos(1)

Documents

261645

Sigma-Aldrich

4-Aminophenethyl alcohol

98%

Synonym(s):

2-(4-Aminophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2CH2OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
907205
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

107-110 °C (lit.)

SMILES string

Nc1ccc(CCO)cc1

InChI

1S/C8H11NO/c9-8-3-1-7(2-4-8)5-6-10/h1-4,10H,5-6,9H2

InChI key

QXHDYMUPPXAMPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Aminophenethyl alcohol has been used:
  • as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester)
  • in the synthesis of 4-aminostyrene
  • in functionalization of graphene nanoplatelets

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constitutional isomerism IV. Synthesis and characterization of poly (amide-ester) s from isophthaloyl chloride and 4-aminophenethyl alcohol.
Li L, et al.
Polymer Journal, 33(4), 364-370 (2001)
Functionalized graphene nanoplatelets for enhanced mechanical and thermal properties of polyurethane nanocomposites.
Yadav SK and Cho JW.
Applied Surface Science, 266, 360-367 (2013)
Synthesis and polymerization of N-(4-vinylphenyl) maleimide.
Hagiwara T, et al.
Macromolecules, 24(26), 6856-6858 (1991)
Mariusz Sandomierski et al.
Scientific reports, 10(1), 19289-19289 (2020-11-08)
Commonly used endoprostheses in the orthopedic industry are those made of Ti6Al4V titanium alloy. Unfortunately, this material has low abrasion resistance, and therefore methods of their modification are still sought. A sensible approach is coating the alloy with a layer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service