256811
Diisobutylaluminum hydride
reagent grade
Synonym(s):
DIBAL, DIBAL-H
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein:
4123663
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
reagent grade
Quality Level
reaction suitability
reagent type: reductant
bp
116-118 °C/1 mmHg (lit.)
density
0.798 g/mL at 25 °C (lit.)
SMILES string
[H][Al](CC(C)C)CC(C)C
InChI
1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;
InChI key
AZWXAPCAJCYGIA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Diisobutylaluminum hydride is commonly used as a reducing agent in organic synthesis.
Application
Diisobutylaluminum hydride can be used in the following protocols:
- As a promotor of Tishchenko reaction of aldehydes.
- Conversion of benzylidene acetals of 1,2-and 1,3-glycols to the corresponding monobenzyl ethers of the glycols.
- To generate a novel chiral reducing agent based on (S)-proline for the asymmetric reduction of prochiral ketones.
Packaging
Supplied in a Sure/Pac™ cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.
Compatible with the following:
Compatible with the following:
- Aldrich® Sure/Pac™ station for liquefied gases Z566446
- PTFE Sealing tape Z104388 or Z221880
- Straight septum-inlet adapter Z118141 with septa Z565687 or Z565695.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
also commonly purchased with this product
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
septum inlet adapter
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Asymmetric reduction of prochiral keto esters with a chiral reducing agent prepared from tin (II) chloride, chiral diamine, and diisobutylaluminum hydride.
Mukaiyama T, et al.
Chemistry Letters (Jpn), 14(6), 813-816 (1985)
Tishchenko reactions of aldehydes promoted by diisobutylaluminum hydride and its application to the macrocyclic lactone formation.
Hon YS, et al.
Tetrahedron, 63(46), 11325-11340 (2007)
A facile cleavage of benzylidene acetals with diisobutylaluminum hydride.
Takano S, et al.
Chemistry Letters (Jpn), 12(10), 1593-1596 (1983)
Daisuke Ando et al.
Journal of agricultural and food chemistry, 66(39), 10154-10162 (2018-09-13)
The metabolic fate of a new fungicide, mandestrobin, labeled with 14C at the phenoxy or benzyl ring was examined in wheat after a single spray application at 300 g/ha. Mandestrobin penetrated into foliage over time, with both radiolabels showing similar
Reaction of diisobutylaluminum hydride with selected organic compounds containing representative functional groups
Yoon N M
The Journal of Organic Chemistry, 50(14), 2443-2450 (1985)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service