Skip to Content
Merck
All Photos(2)

Key Documents

255963

Sigma-Aldrich

Aluminum phosphate

99.99% trace metals basis

Synonym(s):

Aluminum monophosphate, Aluminum orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlO4P
CAS Number:
Molecular Weight:
121.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
(Powder or Crystal or Chunk(s))

Quality Level

Assay

99.99% trace metals basis

form

(Powder or Crystal or Chunk(s))

impurities

≤150.0 ppm Trace Metal Analysis

density

2.56 g/mL at 25 °C (lit.)

SMILES string

[Al+3].[O-]P([O-])([O-])=O

InChI

1S/Al.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3

InChI key

ILRRQNADMUWWFW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum phosphate is a white crystalline solid with excellent thermal stability and a wide bandgap. It also possesses high dielectric constant and insulation properties. It is widely used in the field of solar cells, capacitors, and fuel cells.

Application

Aluminum phosphate can be used:
  • As a coating material to modify the surface of NCM cathode of Li-ion batteries. It helps to enhance the discharge capacity and capacity retention ability of batteries.
  • As a bifunctional additive to fabricate electrode for Li-S batteries. It acts as a lithium polysulfide capturing agent to reduce and stabilize interfacial resistance during cycling.
  • To fabricate polypropylene separator with low ionic impedances and high thermal/electrochemical stability. This leads to improved ionic conductivity and Li-ion diffusion coefficient.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenlei Xie et al.
Bioresource technology, 119, 60-65 (2012-06-26)
WO(3)/AlPO(4) catalysts were prepared by impregnation of AlPO(4) with ammonium metatungstate. Powder X-ray diffraction (XRD), Fourier transform infrared (FT-IR) spectroscopy, and thermo gravimetric and differential thermal analysis (TG-DTA) demonstrated that the tungsten compound was incorporated into AlPO(4) forming the catalyst
Fikru Tafesse et al.
Talanta, 83(5), 1491-1495 (2011-01-18)
A highly selective and sensitive phosphate sensor has been fabricated by constructing a crystal disk consisting of variable mixtures of aluminium powder (Al), aluminium phosphate (AlPO(4)) and powdered copper (Cu). The membrane sensor exhibits linear potential response in the concentration
Belma Ljutic et al.
Vaccine, 30(19), 2981-2988 (2012-03-03)
We investigated the immunogenicity, stability and adsorption properties of an experimental pneumococcal vaccine composed of three protein vaccine antigens; Pneumococcal histidine triad protein D, (PhtD), Pneumococcal choline-binding protein A (PcpA) and genetically detoxified pneumolysin D1 (PlyD1) formulated with aluminum salt
Shibo Jiang et al.
Expert review of vaccines, 11(12), 1405-1413 (2012-12-21)
A subunit vaccine, RBD-S, is under development to prevent severe acute respiratory syndrome (SARS) caused by SARS coronavirus (SARS-CoV), which is classified by the US NIH as a category C pathogen. This vaccine is comprised of a recombinant receptor-binding domain
Ai-Hsiang Chou et al.
PloS one, 7(4), e34834-e34834 (2012-04-25)
Enterovirus 71 (EV71) has caused several epidemics of hand, foot and mouth diseases (HFMD) in Asia and now is being recognized as an important neurotropic virus. Effective medications and prophylactic vaccine against EV71 infection are urgently needed. Based on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service