Skip to Content
Merck
All Photos(1)

Key Documents

254754

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine manganese(III) chloride

95%

Synonym(s):

meso-Tetraphenylporphyrin manganese(III) chloride complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28ClMnN4
CAS Number:
Molecular Weight:
703.11
Beilstein:
5699425
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
core: manganese

λmax

475 nm
583 nm (2nd)

SMILES string

Cl[Mn]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28N4.ClH.Mn/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;

InChI key

MIUMWNDEIWVIEG-YKKPBKTHSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang Xiao-wei et al.
Food chemistry, 197(Pt A), 930-936 (2015-12-01)
Trimethylamine (TMA) is a key measurement indicator for meat spoilage. In order to develop simple, cheap, and sensitive sensors for TMA detection, a nanoporous colorimetric sensor array (NCSA) was developed. A sol-gel method has been used to obtain TiO2 nanoporous
Sumana Kladsomboon et al.
Sensors (Basel, Switzerland), 18(10) (2018-09-23)
Hybrid optical gas sensors, based on different organic and inorganic materials, are proposed in this paper, with the aim of using them as optical artificial nose systems. Three types of organic and inorganic dyes, namely zinc-porphyrin, manganese-porphyrin, and zinc-phthalocyanine, were
B Cheng et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 2), 361-363 (1996-02-15)
The crystal structure of [Mn(TPP)Cl] in space group I4/m (where TPP is C44H28N4) has been determined. The unit cell contains two full molecules, with one eighth of a molecule unique. An out-of-plane model for the Mn atom was applied and
J Krzystek et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(6), 1113-1127 (2002-05-08)
High-field and -frequency electron paramagnetic resonance (HFEPR) spectroscopy has been used to study three complexes of high spin Manganese(III), 3d4, S = 2. The complexes studied were tetraphenylporphyrinatomanganese(III) chloride (MnTPPCI), phthalocyanatomanganese(III) chloride (MnPcCl), and (8,12-diethyl-2,3,7,13,17,18-hexamethylcorrolato)manganese(III) (MnCor). We demonstrate the ability
Majid Moghadam et al.
Bioorganic & medicinal chemistry letters, 16(7), 2026-2030 (2006-01-24)
Mild and efficient oxidation of Hantzsch 1,4-dihydropyridines with sodium periodate catalyzed by Mn(TTP)Cl supported on polystyrene-bound imidazole is reported. This heterogeneous catalyst is of great stability and reusability in the oxidation of 1,4-dihydropyridines with sodium periodate without significant loss of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service