Skip to Content
Merck
All Photos(1)

Documents

254339

Sigma-Aldrich

2-Butyne

99%

Synonym(s):

Crotonylene, Dimethylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CCH3
CAS Number:
Molecular Weight:
54.09
Beilstein:
1361340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

11.47 psi ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.393 (lit.)

bp

27 °C (lit.)

mp

−32 °C (lit.)

density

0.691 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC#CC

InChI

1S/C4H6/c1-3-4-2/h1-2H3

InChI key

XNMQEEKYCVKGBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The ion-molecule reaction of the aromatic distonic N-methyl-pyridinium-4-yl radical cation with 2-butyne has been investigated using ion trap mass spectrometry.

Application

2-Butyne has been used in preparation of THF-free metallacyclobutene complexes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C J Kees-Jan Weststrate et al.
Nature communications, 11(1), 750-750 (2020-02-08)
Facile C-C bond formation is essential to the formation of long hydrocarbon chains in Fischer-Tropsch synthesis. Various chain growth mechanisms have been proposed previously, but spectroscopic identification of surface intermediates involved in C-C bond formation is scarce. We here show
Living polymerization of 2-butyne using a well-characterized tantalum catalyst.
Wallace KC, et al.
Organometallics, 8(3), 644-654 (1989)
Andrea Cernuto et al.
The Journal of chemical physics, 147(15), 154302-154302 (2017-10-23)
The methyl carbocation is ubiquitous in gaseous environments, such as planetary ionospheres, cometary comae, and the interstellar medium, as well as combustion systems and plasma setups for technological applications. Here we report on a joint experimental and theoretical study on
A K Y Lam et al.
Physical chemistry chemical physics : PCCP, 14(7), 2417-2426 (2012-01-18)
Aromatic radicals form in a variety of reacting gas-phase systems, where their molecular weight growth reactions with unsaturated hydrocarbons are of considerable importance. We have investigated the ion-molecule reaction of the aromatic distonic N-methyl-pyridinium-4-yl (NMP) radical cation with 2-butyne (CH(3)C≡CCH(3))
Hongzhen Wang et al.
The journal of physical chemistry. A, 119(46), 11313-11319 (2015-10-23)
The state-selective photodissociation of diacetylene (C4H2) was studied in the wavelength range of 127.5-164.4 nm by high-resolution Rydberg H atom time-of-flight spectroscopy measurements. In the wavelength region, two Rydberg series nR and nR' were state-selectively excited using tunable vacuum-ultraviolet laser

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service